Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IMP4
Entrez Gene ID
92856
Gene Name
IMP4 homolog, U3 small nucleolar ribonucleoprotein
Gene Aliases
BXDC4
Chromosome Location
Chr.2: 130342225 - 130347961 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320304.1 NP_001307233.1 6 - 7 1294 77
NM_001320305.1 NP_001307234.1 6 - 7 1246 77
NM_001320306.1 NP_001307235.1 6 - 7 1313 77
NM_001320307.1 NP_001307236.1 6 - 7 1340 77
NM_001320309.1 NP_001307238.1 5 - 6 1450 77
NM_001320310.1 NP_001307239.1 6 - 7 1327 77
NM_001320311.1 NP_001307240.1 6 - 7 1318 77
NM_033416.2 NP_219484.1 6 - 7 1300 77
GenBank mRNA AF054996.1 - 6 - 7 1115 77
AK289878.1 - 6 - 7 628 77
AK292345.1 - 6 - 7 628 77
AK312808.1 - 6 - 7 628 77
AY364245.1 - 5 - 6 592 77
BC010042.1 - 6 - 7 609 77

View Full Product Details