Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EBF1
Entrez Gene ID
1879
Gene Name
early B-cell factor 1
Gene Aliases
COE1, EBF, O/E-1, OLF1
Chromosome Location
Chr.5: 158695915 - 159099762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290360.2 NP_001277289.1 7 - 8 924 64
NM_001324101.1 NP_001311030.1 7 - 8 924 64
NM_001324103.1 NP_001311032.1 7 - 8 924 64
NM_001324106.1 NP_001311035.1 7 - 8 924 64
NM_001324107.1 NP_001311036.1 7 - 8 924 64
NM_001324108.1 NP_001311037.1 7 - 8 924 64
NM_001324109.1 NP_001311038.1 7 - 8 924 64
NM_001324111.1 NP_001311040.1 7 - 8 1019 64
NM_024007.4 NP_076870.1 7 - 8 924 64
NM_182708.2 NP_874367.1 6 - 7 855 64
XM_017009192.1 XP_016864681.1 7 - 8 924 64
XM_017009193.1 XP_016864682.1 7 - 8 912 64
XM_017009194.1 XP_016864683.1 7 - 8 924 64
XM_017009195.1 XP_016864684.1 7 - 8 924 64
XM_017009196.1 XP_016864685.1 7 - 8 1859 64
XM_017009197.1 XP_016864686.1 7 - 8 1365 64
XM_017009198.1 XP_016864687.1 7 - 8 1284 64
XM_017009199.1 XP_016864688.1 2 - 3 1275 64
XM_017009200.1 XP_016864689.1 2 - 3 18411 64
XM_017009201.1 XP_016864690.1 2 - 3 18411 64
XM_017009202.1 XP_016864691.1 7 - 8 924 64
XM_017009203.1 XP_016864692.1 7 - 8 924 64
GenBank mRNA AF208502.1 - 7 - 8 626 64
AK289712.1 - 7 - 8 911 64
AK303698.1 - 5 - 6 550 64
AK304438.1 - 7 - 8 627 64
BC038805.1 - 7 - 8 935 64
BC041178.1 - 6 - 7 782 64

View Full Product Details