Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148985828-148986311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3TC2
Entrez Gene ID
79628
Gene Name
SH3 domain and tetratricopeptide repeats 2
Gene Aliases
CMT4C, MNMN
UniGene
Hs.483784
Gene Chromosome Location
Chr.5: 148982150-149063174 on Build GRCh38
Transcripts
NM_024577.3

SNP Detail

SNP ID Chromosome Location
rs373932787 Chr.5: 148985971-148985971 on Build GRCh38
rs929665111 Chr.5: 148986264-148986264 on Build GRCh38
rs922595949 Chr.5: 148986015-148986015 on Build GRCh38
rs916396911 Chr.5: 148986171-148986171 on Build GRCh38
rs894257960 Chr.5: 148986098-148986098 on Build GRCh38
rs960901321 Chr.5: 148986149-148986149 on Build GRCh38
rs1055860820 Chr.5: 148986086-148986086 on Build GRCh38
rs927912301 Chr.5: 148986109-148986109 on Build GRCh38
rs985959368 Chr.5: 148985871-148985871 on Build GRCh38
rs755632071 Chr.5: 148986146-148986146 on Build GRCh38
rs182775401 Chr.5: 148986105-148986105 on Build GRCh38
rs185251230 Chr.5: 148986235-148986235 on Build GRCh38
rs936184277 Chr.5: 148986082-148986082 on Build GRCh38
rs536826583 Chr.5: 148986227-148986228 on Build GRCh38
rs941168782 Chr.5: 148986208-148986208 on Build GRCh38
rs566593704 Chr.5: 148985901-148985901 on Build GRCh38
rs887673778 Chr.5: 148986228-148986228 on Build GRCh38
rs886060101 Chr.5: 148986076-148986076 on Build GRCh38
rs1029916768 Chr.5: 148985898-148985898 on Build GRCh38
rs1018387179 Chr.5: 148985915-148985915 on Build GRCh38
rs891336810 Chr.5: 148985958-148985958 on Build GRCh38
rs946354512 Chr.5: 148986021-148986021 on Build GRCh38
rs566606652 Chr.5: 148985882-148985882 on Build GRCh38
rs994214142 Chr.5: 148985885-148985885 on Build GRCh38
rs548465561 Chr.5: 148985853-148985853 on Build GRCh38
rs749885840 Chr.5: 148986057-148986057 on Build GRCh38
rs990217369 Chr.5: 148986029-148986029 on Build GRCh38
rs145655515 Chr.5: 148986239-148986240 on Build GRCh38
rs926186595 Chr.5: 148986054-148986054 on Build GRCh38
rs918290985 Chr.5: 148986237-148986237 on Build GRCh38
rs1018449656 Chr.5: 148985976-148985976 on Build GRCh38
rs552117989 Chr.5: 148986032-148986032 on Build GRCh38
rs570299170 Chr.5: 148986033-148986033 on Build GRCh38
rs10040598 Chr.5: 148986157-148986157 on Build GRCh38
rs117371981 Chr.5: 148986012-148986012 on Build GRCh38
rs967425272 Chr.5: 148986076-148986076 on Build GRCh38

View Full Product Details