The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004747.3 | NP_004738.3 | 9 - 10 | 1817 | 77 |
XM_005270276.4 | XP_005270333.1 | 9 - 10 | 1771 | 77 | |
XM_006718056.3 | XP_006718119.1 | 9 - 10 | 1780 | 77 | |
XM_006718057.3 | XP_006718120.1 | 9 - 10 | 1787 | 77 | |
XM_011540341.2 | XP_011538643.1 | 9 - 10 | 1941 | 77 | |
XM_011540342.1 | XP_011538644.1 | 9 - 10 | 1519 | 77 | |
XM_011540344.2 | XP_011538646.1 | 8 - 9 | 1426 | 77 | |
XM_011540345.1 | XP_011538647.1 | 8 - 9 | 1551 | 77 | |
XM_011540346.2 | XP_011538648.1 | 9 - 10 | 1786 | 77 | |
XM_011540347.2 | XP_011538649.1 | 5 - 6 | 997 | 77 | |
XM_017016913.1 | XP_016872402.1 | 9 - 10 | 1480 | 77 | |
XM_017016914.1 | XP_016872403.1 | 9 - 10 | 1810 | 77 | |
XM_017016915.1 | XP_016872404.1 | 5 - 6 | 1681 | 77 | |
GenBank mRNA | AB011155.2 | - | 9 - 10 | 1847 | 77 |
AB091676.1 | - | 7 - 8 | 1282 | 77 | |
AK055172.1 | - | 5 - 6 | 735 | 77 | |
AK122931.1 | - | 10 - 11 | 1749 | 77 | |
BC146794.1 | - | 9 - 10 | 1847 | 77 | |
BX649131.1 | - | 5 - 6 | 944 | 77 |