The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199511.2 | NP_955805.1 | 4 - 5 | 2999 | 101 |
NM_199512.2 | NP_955806.1 | 4 - 5 | 2402 | 101 | |
GenBank mRNA | AB052098.1 | - | 4 - 5 | 2616 | 101 |
AB266387.1 | - | 4 - 5 | 3187 | 101 | |
AF506819.1 | - | 4 - 5 | 2316 | 101 | |
AK292881.1 | - | 4 - 5 | 2683 | 101 | |
AK304561.1 | - | 5 - 6 | 1286 | 101 | |
AL833034.1 | - | 4 - 5 | 2270 | 101 | |
AY333429.1 | - | 4 - 5 | 2896 | 101 | |
AY526612.1 | - | 4 - 5 | 2987 | 101 | |
AY548106.1 | - | 4 - 5 | 2972 | 101 | |
AY548107.1 | - | 4 - 5 | 2972 | 101 | |
AY726593.1 | - | 4 - 5 | 2453 | 101 | |
BC073164.1 | - | 3 - 4 | 314 | 101 | |
BC110842.1 | - | 4 - 5 | 2984 | 101 | |
BC116178.1 | - | 4 - 5 | 2889 | 101 | |
BX647117.1 | - | 4 - 5 | 2873 | 101 | |
BX647817.1 | - | 4 - 5 | 2288 | 101 |