The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020830.4 | NP_065881.1 | 4 - 5 | 439 | 116 |
GenBank mRNA | AB037856.1 | - | 4 - 5 | 352 | 116 |
AF411977.1 | - | 4 - 5 | 336 | 116 | |
AJ310568.1 | - | 4 - 5 | 426 | 116 | |
AK022888.1 | - | 4 - 5 | 365 | 116 | |
AK023415.1 | - | 3 - 4 | 233 | 116 | |
BC040525.1 | - | 4 - 5 | 351 | 116 | |
BC065934.1 | - | 4 - 5 | 383 | 116 | |
DA775670.1 | - | 4 - 5 | 439 | 116 |