The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015268.3 | NP_056083.3 | 54 - 55 | 6594 | 72 |
XM_005247245.4 | XP_005247302.1 | 55 - 56 | 6791 | 72 | |
XM_017006036.1 | XP_016861525.1 | 41 - 42 | 5051 | 72 | |
GenBank mRNA | AB014578.2 | - | 54 - 55 | 6643 | 72 |
AK023168.1 | - | 23 - 24 | 3022 | 72 | |
AK125330.1 | - | 31 - 32 | 3978 | 72 | |
AK127112.1 | - | 38 - 39 | 4682 | 72 | |
AY779857.1 | - | 54 - 55 | 6594 | 72 | |
BC000164.3 | - | 7 - 8 | 957 | 72 | |
BC009630.1 | - | 9 - 10 | 1077 | 72 | |
BC040638.2 | - | 4 - 5 | 645 | 72 | |
BC065756.1 | - | 3 - 4 | 353 | 72 | |
BC105580.1 | - | 2 - 3 | 170 | 72 | |
BC151246.1 | - | 55 - 56 | 6664 | 72 |