The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017721.4 | NP_060191.3 | 18 - 19 | 2314 | 74 |
XM_005259972.2 | XP_005260029.1 | 18 - 19 | 2259 | 74 | |
XM_005259973.2 | XP_005260030.1 | 18 - 19 | 2262 | 74 | |
XM_005259974.2 | XP_005260031.1 | 18 - 19 | 2250 | 74 | |
XM_005259975.2 | XP_005260032.1 | 18 - 19 | 2247 | 74 | |
GenBank mRNA | AB097002.1 | - | 18 - 19 | 2256 | 74 |
AF536205.1 | - | 20 - 21 | 2367 | 74 | |
AK000248.1 | - | - | 666 | 74 | |
AK023399.1 | - | 11 - 12 | 1139 | 74 | |
AK026371.1 | - | 2 - 3 | 152 | 74 | |
AK315068.1 | - | 18 - 19 | 2232 | 74 | |
BC006556.2 | - | 13 - 14 | 1418 | 74 | |
BC048345.1 | - | 4 - 5 | 326 | 74 | |
BC064981.1 | - | 18 - 19 | 2218 | 74 |