Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SRGAP1
- Entrez Gene ID
-
57522
- Gene Name
- SLIT-ROBO Rho GTPase activating protein 1
- Gene Aliases
-
ARHGAP13, NMTC2
- Chromosome Location
-
Chr.12: 63844372 - 64147863 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK023899.1 |
- |
1 - 2 |
223 |
91 |
View Full Product Details