The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020531.2 | NP_065392.1 | 5 - 6 | 638 | 68 |
XM_005260763.3 | XP_005260820.1 | 5 - 6 | 779 | 68 | |
GenBank mRNA | AB033767.1 | - | 5 - 6 | 577 | 68 |
AK025834.1 | - | 1 - 2 | 102 | 68 | |
AK026866.1 | - | 2 - 3 | 136 | 68 | |
AK291129.1 | - | 5 - 6 | 599 | 68 | |
AK301959.1 | - | 5 - 6 | 779 | 68 | |
AY359076.1 | - | 4 - 5 | 425 | 68 | |
BC003501.1 | - | 5 - 6 | 562 | 68 | |
CF131759.1 | - | 5 - 6 | 607 | 68 |