Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR92
Entrez Gene ID
116143
Gene Name
WD repeat domain 92
Gene Aliases
-
Chromosome Location
Chr.2: 68130149 - 68157560 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138458.3 NP_612467.1 7 - 8 986 117
GenBank mRNA AK056303.1 - 7 - 8 950 117
AK058090.1 - 7 - 8 859 117
BC020271.1 - 7 - 8 940 117
BC066657.1 - 7 - 8 936 117

View Full Product Details