Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41611112-41611606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT16
Entrez Gene ID
3868
Gene Name
keratin 16
Gene Aliases
CK16, FNEPPK, K16, K1CP, KRT16A, NEPPK, PC1
UniGene
Hs.655160
Gene Chromosome Location
Chr.17: 41609778-41612827 on Build GRCh38
Transcripts
NM_005557.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6697862 c.814G>T p.V272L Chr.17: 41611188-41611188 on Build GRCh38
1710285 c.673C>T p.R225W Chr.17: 41611443-41611443 on Build GRCh38
1710284 c.868C>T p.R290C Chr.17: 41611134-41611134 on Build GRCh38
1710283 c.880G>A p.E294K Chr.17: 41611122-41611122 on Build GRCh38
379136 c.784C>G p.L262V Chr.17: 41611218-41611218 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572592608 Chr.17: 41611461-41611461 on Build GRCh38
rs189594149 Chr.17: 41611249-41611249 on Build GRCh38
rs143374360 Chr.17: 41611484-41611484 on Build GRCh38
rs868417520 Chr.17: 41611518-41611518 on Build GRCh38
rs202223973 Chr.17: 41611152-41611152 on Build GRCh38
rs200894937 Chr.17: 41611253-41611253 on Build GRCh38
rs180738664 Chr.17: 41611323-41611323 on Build GRCh38
rs138889840 Chr.17: 41611417-41611417 on Build GRCh38
rs747700643 Chr.17: 41611531-41611531 on Build GRCh38
rs141302209 Chr.17: 41611180-41611180 on Build GRCh38
rs200270322 Chr.17: 41611329-41611329 on Build GRCh38
rs949776540 Chr.17: 41611148-41611148 on Build GRCh38
rs780467068 Chr.17: 41611247-41611247 on Build GRCh38
rs781520782 Chr.17: 41611429-41611429 on Build GRCh38
rs372994057 Chr.17: 41611235-41611235 on Build GRCh38
rs554877931 Chr.17: 41611164-41611164 on Build GRCh38
rs757825274 Chr.17: 41611426-41611426 on Build GRCh38
rs996015875 Chr.17: 41611517-41611517 on Build GRCh38
rs767939342 Chr.17: 41611169-41611169 on Build GRCh38
rs775511623 Chr.17: 41611252-41611252 on Build GRCh38
rs537418366 Chr.17: 41611405-41611405 on Build GRCh38
rs35882482 Chr.17: 41611532-41611533 on Build GRCh38
rs142812826 Chr.17: 41611442-41611442 on Build GRCh38
rs372759162 Chr.17: 41611445-41611445 on Build GRCh38
rs778861086 Chr.17: 41611136-41611136 on Build GRCh38
rs577984211 Chr.17: 41611506-41611506 on Build GRCh38
rs369686494 Chr.17: 41611324-41611324 on Build GRCh38
rs980933287 Chr.17: 41611558-41611558 on Build GRCh38
rs751692927 Chr.17: 41611496-41611496 on Build GRCh38
rs778190817 Chr.17: 41611420-41611420 on Build GRCh38
rs762344504 Chr.17: 41611497-41611497 on Build GRCh38
rs200123727 Chr.17: 41611201-41611201 on Build GRCh38
rs746207767 Chr.17: 41611516-41611516 on Build GRCh38
rs753409964 Chr.17: 41611523-41611523 on Build GRCh38
rs374847388 Chr.17: 41611526-41611526 on Build GRCh38
rs747321787 Chr.17: 41611350-41611350 on Build GRCh38
rs1026298770 Chr.17: 41611322-41611322 on Build GRCh38
rs147423442 Chr.17: 41611472-41611472 on Build GRCh38
rs201633520 Chr.17: 41611489-41611489 on Build GRCh38
rs759181895 Chr.17: 41611361-41611361 on Build GRCh38
rs374969442 Chr.17: 41611205-41611205 on Build GRCh38
rs763953807 Chr.17: 41611306-41611306 on Build GRCh38
rs774954710 Chr.17: 41611167-41611167 on Build GRCh38
rs139769896 Chr.17: 41611583-41611583 on Build GRCh38
rs148173278 Chr.17: 41611479-41611479 on Build GRCh38
rs376071668 Chr.17: 41611304-41611304 on Build GRCh38
rs367929415 Chr.17: 41611404-41611404 on Build GRCh38
rs372618636 Chr.17: 41611175-41611175 on Build GRCh38
rs370377321 Chr.17: 41611389-41611389 on Build GRCh38
rs145075215 Chr.17: 41611351-41611351 on Build GRCh38
rs776284357 Chr.17: 41611281-41611281 on Build GRCh38
rs754126224 Chr.17: 41611336-41611336 on Build GRCh38
rs773603541 Chr.17: 41611255-41611255 on Build GRCh38
rs560786058 Chr.17: 41611586-41611586 on Build GRCh38
rs769393410 Chr.17: 41611369-41611369 on Build GRCh38
rs776196497 Chr.17: 41611354-41611354 on Build GRCh38
rs773515074 Chr.17: 41611172-41611172 on Build GRCh38
rs768491054 Chr.17: 41611529-41611529 on Build GRCh38
rs765449152 Chr.17: 41611483-41611483 on Build GRCh38
rs141914758 Chr.17: 41611177-41611177 on Build GRCh38
rs760306435 Chr.17: 41611173-41611173 on Build GRCh38
rs370538173 Chr.17: 41611423-41611423 on Build GRCh38
rs779035955 Chr.17: 41611340-41611340 on Build GRCh38
rs554718723 Chr.17: 41611478-41611478 on Build GRCh38
rs752211714 Chr.17: 41611310-41611310 on Build GRCh38
rs772511645 Chr.17: 41611254-41611254 on Build GRCh38
rs758059131 Chr.17: 41611204-41611204 on Build GRCh38
rs758429689 Chr.17: 41611419-41611419 on Build GRCh38
rs753312856 Chr.17: 41611198-41611198 on Build GRCh38
rs762939186 Chr.17: 41611318-41611318 on Build GRCh38
rs146562738 Chr.17: 41611151-41611151 on Build GRCh38
rs201618706 Chr.17: 41611393-41611393 on Build GRCh38
rs773206968 Chr.17: 41611344-41611344 on Build GRCh38
rs111815971 Chr.17: 41611233-41611233 on Build GRCh38
rs761066481 Chr.17: 41611258-41611258 on Build GRCh38
rs368580381 Chr.17: 41611165-41611165 on Build GRCh38
rs201610693 Chr.17: 41611299-41611299 on Build GRCh38
rs373296526 Chr.17: 41611370-41611370 on Build GRCh38
rs753404521 Chr.17: 41611134-41611134 on Build GRCh38
rs561324843 Chr.17: 41611144-41611144 on Build GRCh38
rs377070153 Chr.17: 41611392-41611392 on Build GRCh38
rs761840536 Chr.17: 41611463-41611463 on Build GRCh38
rs551589197 Chr.17: 41611278-41611278 on Build GRCh38
rs113890856 Chr.17: 41611342-41611342 on Build GRCh38
rs553943127 Chr.17: 41611459-41611459 on Build GRCh38
rs565316062 Chr.17: 41611499-41611499 on Build GRCh38
rs768880927 Chr.17: 41611251-41611251 on Build GRCh38
rs772092665 Chr.17: 41611542-41611542 on Build GRCh38
rs770211924 Chr.17: 41611153-41611153 on Build GRCh38
rs113973409 Chr.17: 41611556-41611556 on Build GRCh38
rs548761158 Chr.17: 41611341-41611341 on Build GRCh38
rs555754233 Chr.17: 41611408-41611408 on Build GRCh38
rs778174614 Chr.17: 41611347-41611347 on Build GRCh38
rs755204698 Chr.17: 41611412-41611412 on Build GRCh38
rs770015573 Chr.17: 41611340-41611349 on Build GRCh38
rs751488340 Chr.17: 41611538-41611546 on Build GRCh38
rs2100509 Chr.17: 41611584-41611584 on Build GRCh38
rs763012693 Chr.17: 41611389-41611395 on Build GRCh38
rs754948306 Chr.17: 41611337-41611337 on Build GRCh38
rs774364736 Chr.17: 41611462-41611462 on Build GRCh38
rs4796681 Chr.17: 41611492-41611492 on Build GRCh38
rs752688464 Chr.17: 41611203-41611203 on Build GRCh38
rs776890659 Chr.17: 41611212-41611213 on Build GRCh38
rs368180635 Chr.17: 41611443-41611443 on Build GRCh38
rs758986017 Chr.17: 41611295-41611295 on Build GRCh38

View Full Product Details