Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MBD6
Entrez Gene ID
114785
Gene Name
methyl-CpG binding domain protein 6
Gene Aliases
-
Chromosome Location
Chr.12: 57520710 - 57530148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052897.3 NP_443129.3 7 - 8 2309 82
XM_005268617.1 XP_005268674.1 7 - 8 2362 82
XM_005268618.1 XP_005268675.1 7 - 8 2458 82
XM_005268620.2 XP_005268677.1 7 - 8 2362 82
XM_006719217.1 XP_006719280.1 7 - 8 2207 82
XM_006719218.1 XP_006719281.1 7 - 8 2195 82
XM_011537851.2 XP_011536153.1 7 - 8 3195 82
GenBank mRNA AB067474.1 - 4 - 5 1978 82
AK056399.1 - 7 - 8 2343 82
AK093078.1 - 2 - 3 1204 82
AL834230.1 - 1 - 2 588 82
BC065530.1 - 7 - 8 2309 82

View Full Product Details