The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198566.3 | NP_940968.1 | 6 - 7 | 1550 | 132 |
XM_011514038.2 | XP_011512340.1 | 6 - 7 | 1459 | 132 | |
XM_017009445.1 | XP_016864934.1 | 5 - 6 | 1581 | 132 | |
XM_017009446.1 | XP_016864935.1 | 4 - 5 | 1491 | 132 | |
GenBank mRNA | AK056925.1 | - | 6 - 7 | 1465 | 132 |
BC036867.1 | - | 6 - 7 | 1413 | 132 |