Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 65224665-65225178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK4
Entrez Gene ID
205
Gene Name
adenylate kinase 4
Gene Aliases
AK 4, AK3, AK3L1, AK3L2
UniGene
Hs.10862
Gene Chromosome Location
Chr.1: 65147342-65232145 on Build GRCh38
Transcripts
NM_001005353.2 NM_013410.3 NM_203464.2 XM_017000612.1 XM_017000613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4435412 c.465_466insA p.L156fs*7 Chr.1: 65224778-65224779 on Build GRCh38
911581 c.492C>T p.P164P Chr.1: 65224805-65224805 on Build GRCh38
5091539 c.451G>A p.V151I Chr.1: 65224764-65224764 on Build GRCh38
3419388 c.510G>T p.R170S Chr.1: 65224823-65224823 on Build GRCh38
5161093 c.529G>A p.V177M Chr.1: 65224842-65224842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139919714 Chr.1: 65225149-65225149 on Build GRCh38
rs768280167 Chr.1: 65224841-65224841 on Build GRCh38
rs562410622 Chr.1: 65225089-65225090 on Build GRCh38
rs769701643 Chr.1: 65224721-65224721 on Build GRCh38
rs760091829 Chr.1: 65224878-65224878 on Build GRCh38
rs961028723 Chr.1: 65224934-65224934 on Build GRCh38
rs774119059 Chr.1: 65224743-65224743 on Build GRCh38
rs948858046 Chr.1: 65225010-65225010 on Build GRCh38
rs769724703 Chr.1: 65224747-65224747 on Build GRCh38
rs538519457 Chr.1: 65225104-65225104 on Build GRCh38
rs747627514 Chr.1: 65224850-65224850 on Build GRCh38
rs760952693 Chr.1: 65224752-65224752 on Build GRCh38
rs765779508 Chr.1: 65224803-65224803 on Build GRCh38
rs999373241 Chr.1: 65225115-65225115 on Build GRCh38
rs767979322 Chr.1: 65224789-65224789 on Build GRCh38
rs181170632 Chr.1: 65224708-65224708 on Build GRCh38
rs566955591 Chr.1: 65224990-65224990 on Build GRCh38
rs761052678 Chr.1: 65224870-65224870 on Build GRCh38
rs893281300 Chr.1: 65224735-65224735 on Build GRCh38
rs765745506 Chr.1: 65224882-65224882 on Build GRCh38
rs761596339 Chr.1: 65224744-65224744 on Build GRCh38
rs952551362 Chr.1: 65224967-65224967 on Build GRCh38
rs185251178 Chr.1: 65224806-65224806 on Build GRCh38
rs772771023 Chr.1: 65224854-65224854 on Build GRCh38
rs972588604 Chr.1: 65224940-65224940 on Build GRCh38
rs771693162 Chr.1: 65224853-65224853 on Build GRCh38
rs764324408 Chr.1: 65224915-65224915 on Build GRCh38
rs1026870220 Chr.1: 65224941-65224941 on Build GRCh38
rs199651703 Chr.1: 65224972-65224972 on Build GRCh38
rs749275744 Chr.1: 65224829-65224829 on Build GRCh38
rs371300792 Chr.1: 65224748-65224748 on Build GRCh38
rs375913305 Chr.1: 65225001-65225001 on Build GRCh38
rs915991613 Chr.1: 65225009-65225009 on Build GRCh38
rs745661693 Chr.1: 65224718-65224718 on Build GRCh38
rs375331767 Chr.1: 65225018-65225018 on Build GRCh38
rs765366640 Chr.1: 65224764-65224764 on Build GRCh38
rs751132635 Chr.1: 65224921-65224921 on Build GRCh38
rs1053373165 Chr.1: 65224701-65224701 on Build GRCh38
rs150581248 Chr.1: 65225055-65225055 on Build GRCh38
rs140087070 Chr.1: 65224972-65224973 on Build GRCh38
rs894446097 Chr.1: 65224885-65224885 on Build GRCh38
rs189529023 Chr.1: 65224873-65224873 on Build GRCh38
rs778627421 Chr.1: 65224842-65224842 on Build GRCh38
rs4915685 Chr.1: 65224778-65224778 on Build GRCh38
rs373568860 Chr.1: 65224714-65224714 on Build GRCh38
rs934662608 Chr.1: 65225072-65225072 on Build GRCh38
rs34248479 Chr.1: 65224763-65224763 on Build GRCh38
rs768327450 Chr.1: 65224730-65224730 on Build GRCh38
rs909069409 Chr.1: 65224973-65224983 on Build GRCh38
rs750189239 Chr.1: 65224805-65224805 on Build GRCh38
rs972247685 Chr.1: 65224832-65224832 on Build GRCh38
rs780113793 Chr.1: 65224820-65224820 on Build GRCh38
rs775572547 Chr.1: 65224893-65224893 on Build GRCh38
rs200227504 Chr.1: 65224710-65224710 on Build GRCh38
rs376714987 Chr.1: 65224722-65224722 on Build GRCh38
rs531228657 Chr.1: 65224777-65224777 on Build GRCh38
rs879494043 Chr.1: 65224954-65224954 on Build GRCh38
rs762970903 Chr.1: 65224905-65224905 on Build GRCh38
rs547087895 Chr.1: 65224877-65224877 on Build GRCh38
rs141794523 Chr.1: 65225029-65225029 on Build GRCh38
rs754367675 Chr.1: 65224753-65224753 on Build GRCh38
rs200572843 Chr.1: 65224979-65224980 on Build GRCh38
rs893417168 Chr.1: 65225131-65225131 on Build GRCh38
rs777861246 Chr.1: 65224784-65224784 on Build GRCh38
rs567923618 Chr.1: 65225056-65225056 on Build GRCh38

View Full Product Details