Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145960446-145959938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD34A
Entrez Gene ID
284615
Gene Name
ankyrin repeat domain 34A
Gene Aliases
ANKRD34
UniGene
Hs.620591
Gene Chromosome Location
Chr.1: 145959441-145964582 on Build GRCh38
Transcripts
NM_001039888.3 XM_005277431.4

Gene Symbol
POLR3GL
Entrez Gene ID
84265
Gene Name
RNA polymerase III subunit G like
Gene Aliases
RPC32HOM, flj32422
UniGene
Hs.591456
Gene Chromosome Location
Chr.1: 145959037-145978848 on Build GRCh38
Transcripts
NM_001039888.3 XM_005277431.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4661866 c.1445_1446insG p.F485fs*>13 Chr.1: 145960314-145960315 on Build GRCh38
6600678 c.1438C>T p.R480C Chr.1: 145960322-145960322 on Build GRCh38
7355671 c.1372G>A p.A458T Chr.1: 145960388-145960388 on Build GRCh38
1742767 c.1419C>T p.S473S Chr.1: 145960341-145960341 on Build GRCh38
4925695 c.1403T>A p.L468* Chr.1: 145960357-145960357 on Build GRCh38
View More

View Full Product Details