The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317950.1 | NP_001304879.1 | 8 - 9 | 2129 | 109 |
NM_001317952.1 | NP_001304881.1 | 7 - 8 | 1687 | 109 | |
NM_030767.5 | NP_110394.3 | 8 - 9 | 2074 | 109 | |
XM_005252244.2 | XP_005252301.1 | 10 - 11 | 2246 | 109 | |
XM_005252245.1 | XP_005252302.1 | 9 - 10 | 2104 | 109 | |
XM_005252247.4 | XP_005252304.1 | 8 - 9 | 2125 | 109 | |
XM_005252248.2 | XP_005252305.1 | 8 - 9 | 2242 | 109 | |
XM_006717294.1 | XP_006717357.1 | 8 - 9 | 2058 | 109 | |
XM_006717295.2 | XP_006717358.1 | 8 - 9 | 2242 | 109 | |
XM_011519063.2 | XP_011517365.1 | 7 - 8 | 1737 | 109 | |
XM_011519064.2 | XP_011517366.1 | 8 - 9 | 2242 | 109 | |
XM_011519065.2 | XP_011517367.2 | 8 - 9 | 1717 | 109 | |
XM_011519066.2 | XP_011517368.1 | 8 - 9 | 2242 | 109 | |
XM_017015172.1 | XP_016870661.1 | 9 - 10 | 1844 | 109 | |
GenBank mRNA | AB075848.1 | - | 8 - 9 | 2247 | 109 |
AK024431.1 | - | 6 - 7 | 1022 | 109 | |
AK074040.1 | - | 7 - 8 | 1117 | 109 | |
AK131082.2 | - | 8 - 9 | 2129 | 109 | |
AK160382.1 | - | 8 - 9 | 2129 | 109 | |
AL833220.1 | - | 5 - 6 | 551 | 109 | |
AY703039.1 | - | 9 - 10 | 1729 | 109 | |
AY703040.1 | - | 7 - 8 | 1687 | 109 | |
AY703041.1 | - | 9 - 10 | 1443 | 109 | |
AY703042.1 | - | 7 - 8 | 1256 | 109 | |
AY703043.1 | - | 7 - 8 | 2047 | 109 | |
AY703044.1 | - | 6 - 7 | 1741 | 109 | |
BC055285.1 | - | 8 - 9 | 2074 | 109 |