The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152550.3 | NP_689763.3 | 2 - 3 | 618 | 68 |
XM_005268380.2 | XP_005268437.1 | 2 - 3 | 614 | 68 | |
XM_006714757.1 | XP_006714820.1 | 2 - 3 | 614 | 68 | |
XM_011537567.2 | XP_011535869.1 | 2 - 3 | 614 | 68 | |
XM_011537568.2 | XP_011535870.1 | 2 - 3 | 608 | 68 | |
XM_011537569.2 | XP_011535871.1 | 2 - 3 | 614 | 68 | |
XM_017009136.1 | XP_016864625.1 | 2 - 3 | 608 | 68 | |
GenBank mRNA | AK074234.1 | - | 2 - 3 | 602 | 68 |
AK292576.1 | - | 2 - 3 | 603 | 68 | |
AL833297.1 | - | 1 - 2 | 432 | 68 | |
BC125106.1 | - | 2 - 3 | 575 | 68 | |
BC125107.1 | - | 2 - 3 | 575 | 68 |