Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH3RF2
Entrez Gene ID
153769
Gene Name
SH3 domain containing ring finger 2
Gene Aliases
HEPP1, POSHER, PPP1R39, RNF158
Chromosome Location
Chr.5: 145936563 - 146081773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152550.3 NP_689763.3 9 - 10 2157 92
XM_005268380.2 XP_005268437.1 9 - 10 2171 92
XM_006714757.1 XP_006714820.1 9 - 10 2153 92
XM_011537567.2 XP_011535869.1 9 - 10 2171 92
XM_011537568.2 XP_011535870.1 9 - 10 2165 92
XM_011537569.2 XP_011535871.1 9 - 10 2153 92
XM_017009136.1 XP_016864625.1 9 - 10 2147 92
GenBank mRNA AK058046.1 - 3 - 4 657 92
AK074234.1 - 9 - 10 2141 92
AK292576.1 - 9 - 10 2142 92
AK302024.1 - 4 - 5 627 92
AL833297.1 - 8 - 9 1971 92
BC031650.2 - 2 - 3 615 92
BC073914.1 - 3 - 4 643 92
BC125106.1 - 9 - 10 2114 92
BC125107.1 - 9 - 10 2114 92
FJ695509.1 - 1 - 2 441 92

View Full Product Details