The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030630.2 | NP_085133.1 | 16 - 17 | 2206 | 147 |
XM_005257226.1 | XP_005257283.1 | 16 - 17 | 2201 | 147 | |
GenBank mRNA | AK074401.1 | - | 16 - 17 | 2134 | 147 |
AK075111.1 | - | 6 - 7 | 731 | 147 | |
AK125514.1 | - | 3 - 4 | 1027 | 147 | |
AK304081.1 | - | 16 - 17 | 2090 | 147 | |
BC032219.1 | - | 12 - 13 | 1504 | 147 | |
BC035372.1 | - | 16 - 17 | 2122 | 147 |