Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 13040676-13041165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM9C
Entrez Gene ID
171484
Gene Name
family with sequence similarity 9 member C
Gene Aliases
TEX39C
UniGene
Hs.667546
Gene Chromosome Location
Chr.X: 13035617-13044798 on Build GRCh38
Transcripts
NM_174901.5 XM_005274460.2 XM_017029321.1

SNP Detail

SNP ID Chromosome Location
rs866622085 Chr.X: 13041046-13041046 on Build GRCh38
rs762844749 Chr.X: 13041043-13041043 on Build GRCh38
rs775561703 Chr.X: 13040824-13040824 on Build GRCh38
rs762290881 Chr.X: 13040860-13040860 on Build GRCh38
rs764167173 Chr.X: 13040743-13040743 on Build GRCh38
rs184424364 Chr.X: 13040945-13040945 on Build GRCh38
rs754286867 Chr.X: 13040746-13040746 on Build GRCh38
rs769444360 Chr.X: 13040807-13040807 on Build GRCh38
rs778260274 Chr.X: 13040939-13040939 on Build GRCh38
rs780803601 Chr.X: 13040966-13040966 on Build GRCh38
rs779659660 Chr.X: 13040845-13040845 on Build GRCh38
rs765817685 Chr.X: 13040758-13040758 on Build GRCh38
rs973943757 Chr.X: 13041133-13041133 on Build GRCh38
rs757558758 Chr.X: 13040756-13040756 on Build GRCh38
rs753836666 Chr.X: 13040766-13040766 on Build GRCh38
rs769507487 Chr.X: 13041032-13041032 on Build GRCh38
rs192161110 Chr.X: 13040768-13040768 on Build GRCh38
rs776687707 Chr.X: 13040852-13040852 on Build GRCh38
rs368965856 Chr.X: 13040870-13040870 on Build GRCh38
rs755744608 Chr.X: 13040797-13040800 on Build GRCh38
rs766932630 Chr.X: 13040909-13040909 on Build GRCh38
rs962580083 Chr.X: 13041068-13041068 on Build GRCh38
rs575475127 Chr.X: 13040967-13040967 on Build GRCh38
rs902496707 Chr.X: 13040769-13040769 on Build GRCh38
rs768637686 Chr.X: 13041044-13041044 on Build GRCh38
rs750779607 Chr.X: 13040891-13040891 on Build GRCh38
rs144437013 Chr.X: 13040792-13040792 on Build GRCh38
rs142548539 Chr.X: 13040815-13040815 on Build GRCh38
rs144346517 Chr.X: 13041040-13041040 on Build GRCh38
rs779745244 Chr.X: 13040781-13040781 on Build GRCh38
rs751170473 Chr.X: 13040919-13040919 on Build GRCh38
rs746450378 Chr.X: 13040783-13040783 on Build GRCh38
rs148405468 Chr.X: 13040808-13040808 on Build GRCh38
rs758981309 Chr.X: 13040892-13040892 on Build GRCh38
rs764444670 Chr.X: 13040851-13040851 on Build GRCh38
rs750100672 Chr.X: 13040727-13040727 on Build GRCh38
rs754670317 Chr.X: 13040787-13040787 on Build GRCh38
rs771416437 Chr.X: 13040959-13040959 on Build GRCh38
rs202046647 Chr.X: 13040812-13040812 on Build GRCh38
rs150569673 Chr.X: 13040848-13040848 on Build GRCh38
rs755264143 Chr.X: 13040789-13040789 on Build GRCh38
rs189715806 Chr.X: 13040993-13040993 on Build GRCh38

View Full Product Details