Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132725498-132726005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK8
Entrez Gene ID
158067
Gene Name
adenylate kinase 8
Gene Aliases
AK 8, C9orf98, DDX31
UniGene
Hs.421340
Gene Chromosome Location
Chr.9: 132725574-132878811 on Build GRCh38
Transcripts
NM_001317958.1 NM_001317959.1 NM_152572.2 XM_005272169.2 XM_006716965.2 XM_011518278.2 XM_017014308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1314589 c.1313G>A p.R438K Chr.9: 132725815-132725815 on Build GRCh38
3212734 c.1282C>T p.Q428* Chr.9: 132725846-132725846 on Build GRCh38
4970070 c.1399G>A p.E467K Chr.9: 132725729-132725729 on Build GRCh38
753320 c.1270G>T p.D424Y Chr.9: 132725858-132725858 on Build GRCh38
4987079 c.1406G>A p.G469E Chr.9: 132725722-132725722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531715754 Chr.9: 132725536-132725536 on Build GRCh38
rs958370309 Chr.9: 132725521-132725521 on Build GRCh38
rs1048521054 Chr.9: 132725605-132725605 on Build GRCh38
rs1036169119 Chr.9: 132725754-132725754 on Build GRCh38
rs770804748 Chr.9: 132725667-132725667 on Build GRCh38
rs866903749 Chr.9: 132725780-132725780 on Build GRCh38
rs764330683 Chr.9: 132725537-132725537 on Build GRCh38
rs747724625 Chr.9: 132725913-132725913 on Build GRCh38
rs189581069 Chr.9: 132725747-132725747 on Build GRCh38
rs367966120 Chr.9: 132725745-132725745 on Build GRCh38
rs965374543 Chr.9: 132725631-132725631 on Build GRCh38
rs917622287 Chr.9: 132725552-132725552 on Build GRCh38
rs529986751 Chr.9: 132725637-132725637 on Build GRCh38
rs372600462 Chr.9: 132725781-132725781 on Build GRCh38
rs375825588 Chr.9: 132725675-132725675 on Build GRCh38
rs146874552 Chr.9: 132725767-132725767 on Build GRCh38
rs976003191 Chr.9: 132725548-132725548 on Build GRCh38
rs1004334426 Chr.9: 132725627-132725627 on Build GRCh38
rs995686419 Chr.9: 132725574-132725574 on Build GRCh38
rs370791861 Chr.9: 132725685-132725685 on Build GRCh38
rs931896348 Chr.9: 132725554-132725554 on Build GRCh38
rs768062930 Chr.9: 132725704-132725704 on Build GRCh38
rs776143432 Chr.9: 132725949-132725949 on Build GRCh38
rs988355742 Chr.9: 132725973-132725973 on Build GRCh38
rs1000821141 Chr.9: 132725795-132725795 on Build GRCh38
rs949018912 Chr.9: 132725569-132725569 on Build GRCh38
rs533440034 Chr.9: 132725742-132725742 on Build GRCh38
rs996812466 Chr.9: 132725777-132725777 on Build GRCh38
rs760966228 Chr.9: 132725788-132725788 on Build GRCh38
rs758481156 Chr.9: 132725932-132725932 on Build GRCh38
rs750837318 Chr.9: 132725718-132725718 on Build GRCh38
rs77647870 Chr.9: 132725641-132725641 on Build GRCh38
rs536070571 Chr.9: 132725790-132725790 on Build GRCh38
rs184476196 Chr.9: 132725730-132725730 on Build GRCh38
rs778030539 Chr.9: 132725958-132725958 on Build GRCh38
rs572788992 Chr.9: 132725892-132725892 on Build GRCh38
rs1050690053 Chr.9: 132725564-132725564 on Build GRCh38
rs139142699 Chr.9: 132725782-132725782 on Build GRCh38
rs779127805 Chr.9: 132725663-132725663 on Build GRCh38
rs1045362769 Chr.9: 132725570-132725570 on Build GRCh38
rs306524 Chr.9: 132725625-132725625 on Build GRCh38
rs376085711 Chr.9: 132725665-132725665 on Build GRCh38
rs200678908 Chr.9: 132725722-132725722 on Build GRCh38
rs754008893 Chr.9: 132725644-132725644 on Build GRCh38
rs148764112 Chr.9: 132725738-132725738 on Build GRCh38
rs758685979 Chr.9: 132725874-132725874 on Build GRCh38
rs576472974 Chr.9: 132725968-132725968 on Build GRCh38
rs894004339 Chr.9: 132725916-132725916 on Build GRCh38
rs887311396 Chr.9: 132725753-132725753 on Build GRCh38
rs901238251 Chr.9: 132725775-132725775 on Build GRCh38
rs368717209 Chr.9: 132725802-132725802 on Build GRCh38
rs396801 Chr.9: 132725980-132725980 on Build GRCh38
rs868784677 Chr.9: 132725870-132725870 on Build GRCh38
rs944020786 Chr.9: 132725541-132725541 on Build GRCh38
rs897523493 Chr.9: 132725620-132725620 on Build GRCh38
rs773563325 Chr.9: 132725784-132725784 on Build GRCh38
rs764248182 Chr.9: 132725959-132725959 on Build GRCh38
rs772797040 Chr.9: 132725914-132725914 on Build GRCh38
rs144348048 Chr.9: 132725709-132725709 on Build GRCh38
rs28477720 Chr.9: 132725739-132725739 on Build GRCh38
rs550749419 Chr.9: 132725573-132725573 on Build GRCh38
rs1052609389 Chr.9: 132725549-132725549 on Build GRCh38
rs763185252 Chr.9: 132725957-132725957 on Build GRCh38
rs199655656 Chr.9: 132725701-132725701 on Build GRCh38
rs988206339 Chr.9: 132725684-132725684 on Build GRCh38
rs767838160 Chr.9: 132725715-132725715 on Build GRCh38
rs979680875 Chr.9: 132725668-132725668 on Build GRCh38
rs755888977 Chr.9: 132725878-132725878 on Build GRCh38
rs760071529 Chr.9: 132725933-132725933 on Build GRCh38
rs745774866 Chr.9: 132725677-132725677 on Build GRCh38
rs776905012 Chr.9: 132725928-132725928 on Build GRCh38
rs373817929 Chr.9: 132725658-132725658 on Build GRCh38
rs1020103880 Chr.9: 132725630-132725630 on Build GRCh38
rs754920514 Chr.9: 132725887-132725887 on Build GRCh38
rs779703089 Chr.9: 132725746-132725747 on Build GRCh38
rs779977107 Chr.9: 132725639-132725639 on Build GRCh38
rs756005237 Chr.9: 132725745-132725746 on Build GRCh38
rs749218902 Chr.9: 132725885-132725885 on Build GRCh38
rs752974633 Chr.9: 132725808-132725808 on Build GRCh38
rs138440349 Chr.9: 132725700-132725700 on Build GRCh38
rs372487313 Chr.9: 132725756-132725756 on Build GRCh38
rs773892056 Chr.9: 132725771-132725771 on Build GRCh38
rs396819 Chr.9: 132725967-132725967 on Build GRCh38
rs774005338 Chr.9: 132725705-132725705 on Build GRCh38
rs1028243259 Chr.9: 132725778-132725778 on Build GRCh38
rs752879236 Chr.9: 132725858-132725858 on Build GRCh38
rs746666636 Chr.9: 132725925-132725925 on Build GRCh38
rs766978504 Chr.9: 132725729-132725729 on Build GRCh38
rs779744213 Chr.9: 132725879-132725879 on Build GRCh38
rs764342264 Chr.9: 132725876-132725876 on Build GRCh38
rs771290809 Chr.9: 132725927-132725927 on Build GRCh38
rs1024618676 Chr.9: 132725628-132725628 on Build GRCh38
rs1028881427 Chr.9: 132725648-132725648 on Build GRCh38
rs759996983 Chr.9: 132725808-132725808 on Build GRCh38
rs368473946 Chr.9: 132725755-132725755 on Build GRCh38
rs781168820 Chr.9: 132725671-132725671 on Build GRCh38
rs554473389 Chr.9: 132725877-132725877 on Build GRCh38
rs765333161 Chr.9: 132725841-132725841 on Build GRCh38
rs769479139 Chr.9: 132725751-132725751 on Build GRCh38
rs546172258 Chr.9: 132725939-132725939 on Build GRCh38

View Full Product Details