Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153908988-153909492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP4
Entrez Gene ID
393
Gene Name
Rho GTPase activating protein 4
Gene Aliases
C1, RGC1, RhoGAP4, SrGAP4, p115
UniGene
Hs.701324
Gene Chromosome Location
Chr.X: 153907376-153926260 on Build GRCh38
Transcripts
NM_001164741.1 NM_001666.4

SNP Detail

SNP ID Chromosome Location
rs782811401 Chr.X: 153909411-153909411 on Build GRCh38
rs781896813 Chr.X: 153909239-153909239 on Build GRCh38
rs781828828 Chr.X: 153909155-153909155 on Build GRCh38
rs782124817 Chr.X: 153909157-153909157 on Build GRCh38
rs781903759 Chr.X: 153909159-153909159 on Build GRCh38
rs782074399 Chr.X: 153909156-153909156 on Build GRCh38
rs782371290 Chr.X: 153909461-153909461 on Build GRCh38
rs916019364 Chr.X: 153909128-153909128 on Build GRCh38
rs781803925 Chr.X: 153909409-153909409 on Build GRCh38
rs782435401 Chr.X: 153909410-153909410 on Build GRCh38
rs782675835 Chr.X: 153909107-153909107 on Build GRCh38
rs782393297 Chr.X: 153909117-153909117 on Build GRCh38
rs782137662 Chr.X: 153909023-153909023 on Build GRCh38
rs782276308 Chr.X: 153909116-153909116 on Build GRCh38
rs782529644 Chr.X: 153909160-153909160 on Build GRCh38
rs781918744 Chr.X: 153909130-153909130 on Build GRCh38
rs782065792 Chr.X: 153909048-153909048 on Build GRCh38
rs1049311241 Chr.X: 153909205-153909205 on Build GRCh38
rs369331728 Chr.X: 153909186-153909186 on Build GRCh38
rs782335899 Chr.X: 153909201-153909201 on Build GRCh38
rs782020117 Chr.X: 153909020-153909020 on Build GRCh38
rs782168116 Chr.X: 153909393-153909393 on Build GRCh38
rs782786197 Chr.X: 153909190-153909190 on Build GRCh38
rs782430569 Chr.X: 153909443-153909443 on Build GRCh38
rs1046237180 Chr.X: 153909434-153909434 on Build GRCh38
rs781857099 Chr.X: 153909168-153909168 on Build GRCh38
rs34046683 Chr.X: 153909415-153909416 on Build GRCh38
rs782221515 Chr.X: 153909195-153909195 on Build GRCh38
rs781950511 Chr.X: 153909047-153909047 on Build GRCh38
rs782357603 Chr.X: 153909035-153909035 on Build GRCh38
rs149449709 Chr.X: 153909305-153909305 on Build GRCh38
rs781917726 Chr.X: 153909145-153909145 on Build GRCh38
rs782661586 Chr.X: 153909161-153909161 on Build GRCh38
rs1029373622 Chr.X: 153909356-153909356 on Build GRCh38
rs370658465 Chr.X: 153909428-153909428 on Build GRCh38
rs111653090 Chr.X: 153909471-153909471 on Build GRCh38
rs781974499 Chr.X: 153909466-153909466 on Build GRCh38
rs945875701 Chr.X: 153909076-153909076 on Build GRCh38
rs782450081 Chr.X: 153909106-153909106 on Build GRCh38
rs782709109 Chr.X: 153909334-153909334 on Build GRCh38
rs888024184 Chr.X: 153909230-153909230 on Build GRCh38
rs888053601 Chr.X: 153909470-153909470 on Build GRCh38
rs782606987 Chr.X: 153909084-153909084 on Build GRCh38
rs919316738 Chr.X: 153909448-153909448 on Build GRCh38
rs377410859 Chr.X: 153909425-153909425 on Build GRCh38
rs781978537 Chr.X: 153909085-153909085 on Build GRCh38
rs200205320 Chr.X: 153909364-153909364 on Build GRCh38
rs977337947 Chr.X: 153909089-153909089 on Build GRCh38
rs782615973 Chr.X: 153909119-153909119 on Build GRCh38
rs782001991 Chr.X: 153909402-153909402 on Build GRCh38
rs782485822 Chr.X: 153909426-153909426 on Build GRCh38
rs142056530 Chr.X: 153909137-153909137 on Build GRCh38
rs782464835 Chr.X: 153909174-153909174 on Build GRCh38
rs782683862 Chr.X: 153909049-153909049 on Build GRCh38
rs781859806 Chr.X: 153909059-153909059 on Build GRCh38
rs782494446 Chr.X: 153909066-153909066 on Build GRCh38
rs782533260 Chr.X: 153909465-153909465 on Build GRCh38
rs782584354 Chr.X: 153909180-153909180 on Build GRCh38
rs782112340 Chr.X: 153909149-153909149 on Build GRCh38
rs374692439 Chr.X: 153909074-153909074 on Build GRCh38
rs781974611 Chr.X: 153909194-153909194 on Build GRCh38
rs11405331 Chr.X: 153909413-153909414 on Build GRCh38
rs375243502 Chr.X: 153909367-153909368 on Build GRCh38
rs202239294 Chr.X: 153909120-153909120 on Build GRCh38
rs782774727 Chr.X: 153909057-153909057 on Build GRCh38
rs781993268 Chr.X: 153909148-153909148 on Build GRCh38
rs201366489 Chr.X: 153909416-153909416 on Build GRCh38
rs11394527 Chr.X: 153909365-153909366 on Build GRCh38
rs113896893 Chr.X: 153909398-153909398 on Build GRCh38
rs781897609 Chr.X: 153909050-153909050 on Build GRCh38
rs369101749 Chr.X: 153909429-153909429 on Build GRCh38
rs967211052 Chr.X: 153909357-153909357 on Build GRCh38
rs782266940 Chr.X: 153909458-153909458 on Build GRCh38
rs782716842 Chr.X: 153909407-153909407 on Build GRCh38
rs200840051 Chr.X: 153909444-153909444 on Build GRCh38
rs925899872 Chr.X: 153909408-153909408 on Build GRCh38
rs782750206 Chr.X: 153909150-153909150 on Build GRCh38
rs781928234 Chr.X: 153909208-153909208 on Build GRCh38
rs998357342 Chr.X: 153909333-153909333 on Build GRCh38
rs781884568 Chr.X: 153909412-153909412 on Build GRCh38
rs782152456 Chr.X: 153909133-153909133 on Build GRCh38
rs782325624 Chr.X: 153909127-153909127 on Build GRCh38
rs782788360 Chr.X: 153909331-153909331 on Build GRCh38
rs1018340047 Chr.X: 153909307-153909307 on Build GRCh38

View Full Product Details