Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153910838-153911249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP4
Entrez Gene ID
393
Gene Name
Rho GTPase activating protein 4
Gene Aliases
C1, RGC1, RhoGAP4, SrGAP4, p115
UniGene
Hs.701324
Gene Chromosome Location
Chr.X: 153907376-153926260 on Build GRCh38
Transcripts
NM_001164741.1 NM_001666.4

SNP Detail

SNP ID Chromosome Location
rs983166553 Chr.X: 153911002-153911002 on Build GRCh38
rs782704073 Chr.X: 153910905-153910905 on Build GRCh38
rs782046266 Chr.X: 153911182-153911182 on Build GRCh38
rs781833504 Chr.X: 153910998-153910998 on Build GRCh38
rs532716652 Chr.X: 153910929-153910929 on Build GRCh38
rs782219026 Chr.X: 153911202-153911202 on Build GRCh38
rs782131699 Chr.X: 153911208-153911208 on Build GRCh38
rs782379357 Chr.X: 153911084-153911084 on Build GRCh38
rs200424312 Chr.X: 153911146-153911146 on Build GRCh38
rs782268390 Chr.X: 153911062-153911062 on Build GRCh38
rs368557149 Chr.X: 153910861-153910861 on Build GRCh38
rs782629689 Chr.X: 153911097-153911097 on Build GRCh38
rs781960915 Chr.X: 153910858-153910858 on Build GRCh38
rs189317801 Chr.X: 153911107-153911107 on Build GRCh38
rs782434268 Chr.X: 153911185-153911185 on Build GRCh38
rs371442614 Chr.X: 153911010-153911010 on Build GRCh38
rs374565768 Chr.X: 153910856-153910856 on Build GRCh38
rs782128441 Chr.X: 153910896-153910896 on Build GRCh38
rs559771643 Chr.X: 153911004-153911004 on Build GRCh38
rs782013683 Chr.X: 153910887-153910887 on Build GRCh38
rs941901715 Chr.X: 153911030-153911030 on Build GRCh38
rs1051659216 Chr.X: 153911142-153911142 on Build GRCh38
rs782624884 Chr.X: 153910975-153910975 on Build GRCh38
rs782460243 Chr.X: 153911100-153911100 on Build GRCh38
rs782681117 Chr.X: 153910955-153910955 on Build GRCh38
rs781878408 Chr.X: 153910954-153910954 on Build GRCh38
rs781969307 Chr.X: 153910901-153910901 on Build GRCh38
rs782809170 Chr.X: 153910940-153910940 on Build GRCh38
rs782534760 Chr.X: 153910914-153910914 on Build GRCh38
rs782561730 Chr.X: 153911084-153911084 on Build GRCh38
rs1042856 Chr.X: 153910967-153910967 on Build GRCh38
rs782756689 Chr.X: 153910900-153910900 on Build GRCh38
rs373784037 Chr.X: 153911106-153911106 on Build GRCh38
rs782426014 Chr.X: 153910886-153910886 on Build GRCh38
rs112645237 Chr.X: 153911079-153911079 on Build GRCh38
rs979819296 Chr.X: 153910862-153910862 on Build GRCh38
rs973641523 Chr.X: 153911049-153911049 on Build GRCh38
rs782360212 Chr.X: 153910857-153910857 on Build GRCh38
rs935282101 Chr.X: 153911147-153911147 on Build GRCh38
rs2070098 Chr.X: 153911163-153911163 on Build GRCh38
rs931878792 Chr.X: 153911118-153911118 on Build GRCh38

View Full Product Details