Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153912621-153913117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP4
Entrez Gene ID
393
Gene Name
Rho GTPase activating protein 4
Gene Aliases
C1, RGC1, RhoGAP4, SrGAP4, p115
UniGene
Hs.701324
Gene Chromosome Location
Chr.X: 153907376-153926260 on Build GRCh38
Transcripts
NM_001164741.1 NM_001666.4

SNP Detail

SNP ID Chromosome Location
rs781847138 Chr.X: 153912696-153912696 on Build GRCh38
rs199594389 Chr.X: 153912999-153912999 on Build GRCh38
rs910221188 Chr.X: 153912762-153912762 on Build GRCh38
rs781993078 Chr.X: 153912814-153912814 on Build GRCh38
rs543748181 Chr.X: 153912663-153912663 on Build GRCh38
rs782741535 Chr.X: 153913076-153913076 on Build GRCh38
rs186003379 Chr.X: 153912969-153912969 on Build GRCh38
rs200697971 Chr.X: 153912993-153912993 on Build GRCh38
rs782493227 Chr.X: 153912709-153912709 on Build GRCh38
rs782274267 Chr.X: 153912732-153912732 on Build GRCh38
rs782682382 Chr.X: 153912726-153912726 on Build GRCh38
rs375094345 Chr.X: 153912991-153912991 on Build GRCh38
rs781818677 Chr.X: 153912820-153912820 on Build GRCh38
rs373421640 Chr.X: 153912994-153912994 on Build GRCh38
rs34279017 Chr.X: 153912725-153912725 on Build GRCh38
rs781795780 Chr.X: 153912673-153912673 on Build GRCh38
rs201338822 Chr.X: 153912755-153912755 on Build GRCh38
rs932908124 Chr.X: 153912758-153912758 on Build GRCh38
rs377026459 Chr.X: 153912759-153912759 on Build GRCh38
rs782598065 Chr.X: 153912985-153912985 on Build GRCh38
rs1015673244 Chr.X: 153913041-153913041 on Build GRCh38
rs782653044 Chr.X: 153912693-153912693 on Build GRCh38
rs782200499 Chr.X: 153912981-153912982 on Build GRCh38
rs373365831 Chr.X: 153912874-153912875 on Build GRCh38
rs782520133 Chr.X: 153912678-153912678 on Build GRCh38
rs369990769 Chr.X: 153912986-153912986 on Build GRCh38
rs782724377 Chr.X: 153913001-153913001 on Build GRCh38
rs782242658 Chr.X: 153912854-153912854 on Build GRCh38
rs782697256 Chr.X: 153912832-153912832 on Build GRCh38
rs782349092 Chr.X: 153912768-153912768 on Build GRCh38
rs782651582 Chr.X: 153912848-153912848 on Build GRCh38
rs782813876 Chr.X: 153912818-153912818 on Build GRCh38
rs781940094 Chr.X: 153912776-153912776 on Build GRCh38
rs781857374 Chr.X: 153913055-153913055 on Build GRCh38
rs782197574 Chr.X: 153912717-153912717 on Build GRCh38
rs782795134 Chr.X: 153913031-153913031 on Build GRCh38
rs182260926 Chr.X: 153912863-153912863 on Build GRCh38
rs781826200 Chr.X: 153913079-153913079 on Build GRCh38
rs77485258 Chr.X: 153913005-153913006 on Build GRCh38
rs782788316 Chr.X: 153912809-153912809 on Build GRCh38
rs782499177 Chr.X: 153912973-153912973 on Build GRCh38
rs781847882 Chr.X: 153912657-153912657 on Build GRCh38
rs781871982 Chr.X: 153913046-153913046 on Build GRCh38
rs5904376 Chr.X: 153913008-153913009 on Build GRCh38
rs974268010 Chr.X: 153912722-153912722 on Build GRCh38
rs782197916 Chr.X: 153912753-153912753 on Build GRCh38
rs781959232 Chr.X: 153912823-153912823 on Build GRCh38
rs782164967 Chr.X: 153912942-153912942 on Build GRCh38
rs782767840 Chr.X: 153912650-153912650 on Build GRCh38
rs782598414 Chr.X: 153912714-153912714 on Build GRCh38
rs373876709 Chr.X: 153912990-153912990 on Build GRCh38
rs782090153 Chr.X: 153912660-153912660 on Build GRCh38
rs781902670 Chr.X: 153912833-153912833 on Build GRCh38
rs782452953 Chr.X: 153913083-153913083 on Build GRCh38
rs782107711 Chr.X: 153912816-153912816 on Build GRCh38
rs368307774 Chr.X: 153912737-153912737 on Build GRCh38
rs782341916 Chr.X: 153912787-153912787 on Build GRCh38
rs782583986 Chr.X: 153913091-153913091 on Build GRCh38
rs1040179521 Chr.X: 153912812-153912812 on Build GRCh38
rs1043552883 Chr.X: 153912879-153912879 on Build GRCh38
rs782099184 Chr.X: 153912767-153912767 on Build GRCh38

View Full Product Details