Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154437149-154437683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDI1
Entrez Gene ID
2664
Gene Name
GDP dissociation inhibitor 1
Gene Aliases
1A, GDIL, MRX41, MRX48, OPHN2, RABGD1A, RABGDIA, XAP-4
UniGene
Hs.595950
Gene Chromosome Location
Chr.X: 154436913-154443467 on Build GRCh38
Transcripts
NM_001493.2

SNP Detail

SNP ID Chromosome Location
rs927223768 Chr.X: 154437366-154437366 on Build GRCh38
rs782689696 Chr.X: 154437626-154437626 on Build GRCh38
rs183953177 Chr.X: 154437233-154437233 on Build GRCh38
rs782033963 Chr.X: 154437414-154437414 on Build GRCh38
rs782401019 Chr.X: 154437220-154437220 on Build GRCh38
rs781846205 Chr.X: 154437178-154437178 on Build GRCh38
rs992678132 Chr.X: 154437376-154437376 on Build GRCh38
rs782590961 Chr.X: 154437187-154437187 on Build GRCh38
rs1009081584 Chr.X: 154437579-154437579 on Build GRCh38
rs782709055 Chr.X: 154437258-154437258 on Build GRCh38
rs782217354 Chr.X: 154437342-154437342 on Build GRCh38
rs782511407 Chr.X: 154437340-154437340 on Build GRCh38
rs782587106 Chr.X: 154437172-154437172 on Build GRCh38
rs373598827 Chr.X: 154437242-154437242 on Build GRCh38
rs782716051 Chr.X: 154437322-154437322 on Build GRCh38
rs782446954 Chr.X: 154437328-154437328 on Build GRCh38
rs782540971 Chr.X: 154437620-154437620 on Build GRCh38
rs782810997 Chr.X: 154437296-154437296 on Build GRCh38
rs201463367 Chr.X: 154437225-154437225 on Build GRCh38
rs1036816756 Chr.X: 154437447-154437447 on Build GRCh38
rs370426944 Chr.X: 154437326-154437326 on Build GRCh38
rs781866156 Chr.X: 154437304-154437304 on Build GRCh38
rs192115845 Chr.X: 154437193-154437193 on Build GRCh38
rs782286083 Chr.X: 154437339-154437339 on Build GRCh38
rs189578067 Chr.X: 154437448-154437448 on Build GRCh38
rs782405711 Chr.X: 154437389-154437389 on Build GRCh38
rs187704798 Chr.X: 154437443-154437443 on Build GRCh38
rs782639908 Chr.X: 154437341-154437341 on Build GRCh38
rs978440305 Chr.X: 154437354-154437354 on Build GRCh38
rs781921921 Chr.X: 154437172-154437173 on Build GRCh38
rs781814710 Chr.X: 154437616-154437616 on Build GRCh38
rs782065640 Chr.X: 154437343-154437343 on Build GRCh38
rs781923617 Chr.X: 154437347-154437347 on Build GRCh38
rs782554121 Chr.X: 154437212-154437212 on Build GRCh38
rs781939489 Chr.X: 154437276-154437276 on Build GRCh38
rs182606975 Chr.X: 154437514-154437514 on Build GRCh38
rs369912312 Chr.X: 154437607-154437607 on Build GRCh38
rs917058114 Chr.X: 154437435-154437435 on Build GRCh38
rs988932047 Chr.X: 154437278-154437278 on Build GRCh38
rs782556919 Chr.X: 154437204-154437204 on Build GRCh38
rs782149450 Chr.X: 154437290-154437290 on Build GRCh38
rs368181402 Chr.X: 154437312-154437312 on Build GRCh38
rs782438799 Chr.X: 154437186-154437186 on Build GRCh38
rs889391884 Chr.X: 154437545-154437545 on Build GRCh38
rs377447247 Chr.X: 154437209-154437209 on Build GRCh38
rs782210039 Chr.X: 154437205-154437205 on Build GRCh38
rs782423142 Chr.X: 154437210-154437211 on Build GRCh38
rs200697643 Chr.X: 154437500-154437500 on Build GRCh38
rs995102787 Chr.X: 154437183-154437183 on Build GRCh38
rs1019085044 Chr.X: 154437651-154437651 on Build GRCh38
rs782105861 Chr.X: 154437249-154437249 on Build GRCh38
rs937222812 Chr.X: 154437372-154437372 on Build GRCh38
rs782672461 Chr.X: 154437338-154437338 on Build GRCh38
rs782192326 Chr.X: 154437219-154437219 on Build GRCh38
rs781862110 Chr.X: 154437633-154437633 on Build GRCh38

View Full Product Details