Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154438431-154438922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDI1
Entrez Gene ID
2664
Gene Name
GDP dissociation inhibitor 1
Gene Aliases
1A, GDIL, MRX41, MRX48, OPHN2, RABGD1A, RABGDIA, XAP-4
UniGene
Hs.595950
Gene Chromosome Location
Chr.X: 154436913-154443467 on Build GRCh38
Transcripts
NM_001493.2

SNP Detail

SNP ID Chromosome Location
rs11549301 Chr.X: 154438823-154438823 on Build GRCh38
rs962289644 Chr.X: 154438678-154438678 on Build GRCh38
rs782126077 Chr.X: 154438507-154438507 on Build GRCh38
rs782681370 Chr.X: 154438658-154438658 on Build GRCh38
rs781899318 Chr.X: 154438758-154438758 on Build GRCh38
rs200590846 Chr.X: 154438745-154438745 on Build GRCh38
rs782773528 Chr.X: 154438886-154438886 on Build GRCh38
rs376966686 Chr.X: 154438888-154438888 on Build GRCh38
rs782220704 Chr.X: 154438480-154438480 on Build GRCh38
rs782404040 Chr.X: 154438649-154438649 on Build GRCh38
rs782298755 Chr.X: 154438497-154438497 on Build GRCh38
rs782648034 Chr.X: 154438479-154438479 on Build GRCh38
rs782242956 Chr.X: 154438675-154438675 on Build GRCh38
rs781978284 Chr.X: 154438501-154438501 on Build GRCh38
rs782398267 Chr.X: 154438816-154438816 on Build GRCh38
rs782458286 Chr.X: 154438748-154438748 on Build GRCh38
rs782546029 Chr.X: 154438600-154438601 on Build GRCh38
rs782616150 Chr.X: 154438877-154438878 on Build GRCh38
rs782559224 Chr.X: 154438594-154438594 on Build GRCh38
rs782306922 Chr.X: 154438732-154438732 on Build GRCh38
rs376041217 Chr.X: 154438761-154438761 on Build GRCh38
rs373515131 Chr.X: 154438837-154438837 on Build GRCh38
rs781964138 Chr.X: 154438824-154438824 on Build GRCh38
rs782446826 Chr.X: 154438558-154438558 on Build GRCh38
rs782454503 Chr.X: 154438897-154438897 on Build GRCh38
rs782638600 Chr.X: 154438599-154438599 on Build GRCh38
rs4834 Chr.X: 154438830-154438830 on Build GRCh38
rs199816411 Chr.X: 154438798-154438798 on Build GRCh38
rs73638282 Chr.X: 154438737-154438737 on Build GRCh38
rs185552165 Chr.X: 154438512-154438512 on Build GRCh38
rs782717448 Chr.X: 154438537-154438537 on Build GRCh38
rs113696246 Chr.X: 154438762-154438762 on Build GRCh38
rs121434608 Chr.X: 154438819-154438819 on Build GRCh38
rs111927791 Chr.X: 154438522-154438522 on Build GRCh38
rs782612758 Chr.X: 154438500-154438500 on Build GRCh38
rs976071793 Chr.X: 154438751-154438751 on Build GRCh38
rs1013957873 Chr.X: 154438640-154438640 on Build GRCh38
rs368687000 Chr.X: 154438681-154438681 on Build GRCh38
rs782386572 Chr.X: 154438476-154438476 on Build GRCh38
rs375881765 Chr.X: 154438679-154438679 on Build GRCh38
rs372327910 Chr.X: 154438613-154438613 on Build GRCh38
rs782469601 Chr.X: 154438894-154438894 on Build GRCh38
rs782571930 Chr.X: 154438645-154438645 on Build GRCh38
rs781796827 Chr.X: 154438516-154438516 on Build GRCh38
rs782099702 Chr.X: 154438715-154438715 on Build GRCh38
rs782192899 Chr.X: 154438775-154438775 on Build GRCh38
rs782485291 Chr.X: 154438767-154438767 on Build GRCh38
rs371989669 Chr.X: 154438750-154438750 on Build GRCh38
rs202041945 Chr.X: 154438555-154438555 on Build GRCh38
rs782672196 Chr.X: 154438461-154438461 on Build GRCh38
rs782154448 Chr.X: 154438863-154438863 on Build GRCh38
rs782509267 Chr.X: 154438738-154438739 on Build GRCh38
rs1043944144 Chr.X: 154438800-154438800 on Build GRCh38
rs199665936 Chr.X: 154438744-154438744 on Build GRCh38
rs369344595 Chr.X: 154438742-154438742 on Build GRCh38
rs782012957 Chr.X: 154438845-154438845 on Build GRCh38
rs374361918 Chr.X: 154438472-154438472 on Build GRCh38
rs782186938 Chr.X: 154438648-154438648 on Build GRCh38
rs782214223 Chr.X: 154438765-154438765 on Build GRCh38
rs376525601 Chr.X: 154438740-154438740 on Build GRCh38
rs782724567 Chr.X: 154438709-154438709 on Build GRCh38
rs782571802 Chr.X: 154438771-154438771 on Build GRCh38
rs188380786 Chr.X: 154438513-154438513 on Build GRCh38
rs781792126 Chr.X: 154438895-154438895 on Build GRCh38
rs1023960992 Chr.X: 154438670-154438670 on Build GRCh38

View Full Product Details