Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154442027-154442536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDI1
Entrez Gene ID
2664
Gene Name
GDP dissociation inhibitor 1
Gene Aliases
1A, GDIL, MRX41, MRX48, OPHN2, RABGD1A, RABGDIA, XAP-4
UniGene
Hs.595950
Gene Chromosome Location
Chr.X: 154436913-154443467 on Build GRCh38
Transcripts
NM_001493.2

SNP Detail

SNP ID Chromosome Location
rs782030017 Chr.X: 154442100-154442100 on Build GRCh38
rs782263053 Chr.X: 154442384-154442384 on Build GRCh38
rs371655047 Chr.X: 154442374-154442374 on Build GRCh38
rs782029331 Chr.X: 154442512-154442512 on Build GRCh38
rs782789881 Chr.X: 154442331-154442331 on Build GRCh38
rs781830959 Chr.X: 154442492-154442492 on Build GRCh38
rs782440937 Chr.X: 154442236-154442236 on Build GRCh38
rs782548681 Chr.X: 154442509-154442509 on Build GRCh38
rs782086050 Chr.X: 154442107-154442107 on Build GRCh38
rs962752161 Chr.X: 154442157-154442157 on Build GRCh38
rs370706163 Chr.X: 154442145-154442145 on Build GRCh38
rs376363700 Chr.X: 154442092-154442092 on Build GRCh38
rs782787069 Chr.X: 154442367-154442367 on Build GRCh38
rs150955632 Chr.X: 154442197-154442197 on Build GRCh38
rs781969275 Chr.X: 154442326-154442326 on Build GRCh38
rs782338931 Chr.X: 154442446-154442446 on Build GRCh38
rs374368885 Chr.X: 154442267-154442267 on Build GRCh38
rs782356686 Chr.X: 154442440-154442440 on Build GRCh38
rs782752472 Chr.X: 154442488-154442488 on Build GRCh38
rs369952655 Chr.X: 154442072-154442072 on Build GRCh38
rs373502838 Chr.X: 154442090-154442090 on Build GRCh38
rs782018234 Chr.X: 154442280-154442280 on Build GRCh38
rs782598409 Chr.X: 154442091-154442091 on Build GRCh38
rs376618857 Chr.X: 154442103-154442103 on Build GRCh38
rs975406035 Chr.X: 154442264-154442264 on Build GRCh38
rs782549320 Chr.X: 154442357-154442357 on Build GRCh38
rs781846403 Chr.X: 154442121-154442121 on Build GRCh38
rs782655354 Chr.X: 154442397-154442397 on Build GRCh38
rs377631325 Chr.X: 154442268-154442268 on Build GRCh38
rs372121683 Chr.X: 154442086-154442086 on Build GRCh38
rs782559739 Chr.X: 154442453-154442453 on Build GRCh38
rs782232197 Chr.X: 154442241-154442241 on Build GRCh38
rs781819035 Chr.X: 154442183-154442183 on Build GRCh38
rs782556991 Chr.X: 154442369-154442369 on Build GRCh38
rs782010840 Chr.X: 154442467-154442467 on Build GRCh38
rs782713548 Chr.X: 154442061-154442061 on Build GRCh38
rs1010600120 Chr.X: 154442051-154442051 on Build GRCh38
rs140855833 Chr.X: 154442404-154442404 on Build GRCh38
rs782402492 Chr.X: 154442223-154442223 on Build GRCh38
rs370898685 Chr.X: 154442277-154442277 on Build GRCh38
rs782282953 Chr.X: 154442434-154442434 on Build GRCh38
rs782629762 Chr.X: 154442198-154442198 on Build GRCh38
rs781884297 Chr.X: 154442444-154442444 on Build GRCh38
rs782055995 Chr.X: 154442369-154442370 on Build GRCh38
rs782004436 Chr.X: 154442266-154442266 on Build GRCh38
rs782068810 Chr.X: 154442290-154442290 on Build GRCh38
rs368434464 Chr.X: 154442354-154442354 on Build GRCh38
rs987108160 Chr.X: 154442351-154442351 on Build GRCh38
rs782289676 Chr.X: 154442200-154442200 on Build GRCh38
rs141599363 Chr.X: 154442101-154442101 on Build GRCh38
rs934237552 Chr.X: 154442323-154442323 on Build GRCh38
rs368966398 Chr.X: 154442094-154442094 on Build GRCh38
rs782169795 Chr.X: 154442125-154442125 on Build GRCh38
rs398122814 Chr.X: 154442421-154442422 on Build GRCh38
rs368190970 Chr.X: 154442466-154442466 on Build GRCh38
rs782606377 Chr.X: 154442167-154442167 on Build GRCh38
rs782062036 Chr.X: 154442473-154442473 on Build GRCh38
rs782370658 Chr.X: 154442097-154442097 on Build GRCh38
rs185040685 Chr.X: 154442439-154442439 on Build GRCh38
rs201330829 Chr.X: 154442301-154442301 on Build GRCh38
rs782501562 Chr.X: 154442386-154442386 on Build GRCh38
rs983015889 Chr.X: 154442135-154442135 on Build GRCh38
rs782346555 Chr.X: 154442249-154442249 on Build GRCh38
rs200407569 Chr.X: 154442464-154442464 on Build GRCh38
rs782798421 Chr.X: 154442133-154442133 on Build GRCh38
rs782785938 Chr.X: 154442112-154442112 on Build GRCh38
rs782600356 Chr.X: 154442378-154442378 on Build GRCh38

View Full Product Details