Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 6865777-6866285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMELY
Entrez Gene ID
266
Gene Name
amelogenin, Y-linked
Gene Aliases
AMGL, AMGY
UniGene
Hs.1238
Gene Chromosome Location
Chr.Y: 6865918-6874056 on Build GRCh38
Transcripts
NM_001143.1 XM_017030042.1

SNP Detail

SNP ID Chromosome Location
rs765549752 Chr.Y: 6866112-6866112 on Build GRCh38
rs778606082 Chr.Y: 6865915-6865915 on Build GRCh38
rs752855344 Chr.Y: 6866117-6866117 on Build GRCh38
rs1009216970 Chr.Y: 6866011-6866011 on Build GRCh38
rs760519968 Chr.Y: 6866079-6866079 on Build GRCh38
rs1022487423 Chr.Y: 6866015-6866015 on Build GRCh38
rs980899057 Chr.Y: 6866158-6866158 on Build GRCh38
rs761791738 Chr.Y: 6866046-6866046 on Build GRCh38
rs926743333 Chr.Y: 6866227-6866227 on Build GRCh38
rs956545502 Chr.Y: 6865987-6865987 on Build GRCh38
rs765569296 Chr.Y: 6866184-6866184 on Build GRCh38
rs537895023 Chr.Y: 6865855-6865855 on Build GRCh38
rs767451839 Chr.Y: 6866050-6866050 on Build GRCh38
rs773181016 Chr.Y: 6866066-6866066 on Build GRCh38

View Full Product Details