Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 6867945-6868449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMELY
Entrez Gene ID
266
Gene Name
amelogenin, Y-linked
Gene Aliases
AMGL, AMGY
UniGene
Hs.1238
Gene Chromosome Location
Chr.Y: 6865918-6874056 on Build GRCh38
Transcripts
NM_001143.1 XM_017030042.1

SNP Detail

SNP ID Chromosome Location
rs753555850 Chr.Y: 6868406-6868406 on Build GRCh38
rs761863669 Chr.Y: 6868343-6868343 on Build GRCh38
rs747629898 Chr.Y: 6868204-6868204 on Build GRCh38
rs763038450 Chr.Y: 6868324-6868324 on Build GRCh38
rs35815655 Chr.Y: 6868402-6868402 on Build GRCh38
rs752070682 Chr.Y: 6868320-6868321 on Build GRCh38
rs1002385553 Chr.Y: 6868376-6868376 on Build GRCh38
rs757767781 Chr.Y: 6868329-6868330 on Build GRCh38
rs781641924 Chr.Y: 6868105-6868105 on Build GRCh38
rs757725231 Chr.Y: 6868100-6868100 on Build GRCh38
rs768073566 Chr.Y: 6868348-6868348 on Build GRCh38
rs758507846 Chr.Y: 6867991-6867991 on Build GRCh38
rs760609997 Chr.Y: 6868309-6868309 on Build GRCh38
rs768233838 Chr.Y: 6868192-6868192 on Build GRCh38
rs200834952 Chr.Y: 6868316-6868316 on Build GRCh38
rs779825282 Chr.Y: 6868114-6868114 on Build GRCh38
rs773693039 Chr.Y: 6868195-6868195 on Build GRCh38
rs376163078 Chr.Y: 6868218-6868218 on Build GRCh38
rs772180487 Chr.Y: 6868267-6868267 on Build GRCh38
rs961143675 Chr.Y: 6868403-6868403 on Build GRCh38
rs2071394 Chr.Y: 6868113-6868113 on Build GRCh38
rs796614753 Chr.Y: 6868024-6868024 on Build GRCh38
rs751678573 Chr.Y: 6868333-6868333 on Build GRCh38
rs892242156 Chr.Y: 6867971-6867971 on Build GRCh38
rs773265244 Chr.Y: 6868302-6868302 on Build GRCh38
rs200941883 Chr.Y: 6868330-6868330 on Build GRCh38
rs775797197 Chr.Y: 6868321-6868321 on Build GRCh38
rs751650146 Chr.Y: 6868180-6868180 on Build GRCh38
rs750929142 Chr.Y: 6868392-6868392 on Build GRCh38
rs754951520 Chr.Y: 6868088-6868088 on Build GRCh38
rs746281657 Chr.Y: 6868111-6868111 on Build GRCh38
rs373182951 Chr.Y: 6868086-6868086 on Build GRCh38
rs764700007 Chr.Y: 6868150-6868158 on Build GRCh38

View Full Product Details