Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 50802005-50802553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A1CF
Entrez Gene ID
29974
Gene Name
APOBEC1 complementation factor
Gene Aliases
ACF, ACF64, ACF65, APOBEC1CF, ASP
UniGene
Hs.282795
Gene Chromosome Location
Chr.10: 50799409-50885681 on Build GRCh38
Transcripts
NM_001198818.1 NM_001198819.1 NM_001198820.1 NM_014576.3 NM_138932.2 NM_138933.2 XM_005269718.2 XM_005269720.3 XM_011539729.2 XM_011539730.2 XM_017016160.1

SNP Detail

SNP ID Chromosome Location
rs369298308 Chr.10: 50802176-50802176 on Build GRCh38
rs913065485 Chr.10: 50802360-50802360 on Build GRCh38
rs962440791 Chr.10: 50802160-50802160 on Build GRCh38
rs987655285 Chr.10: 50802462-50802462 on Build GRCh38
rs140487683 Chr.10: 50802194-50802194 on Build GRCh38
rs905272701 Chr.10: 50802302-50802302 on Build GRCh38
rs572450377 Chr.10: 50802232-50802232 on Build GRCh38
rs147886350 Chr.10: 50802109-50802109 on Build GRCh38
rs547003448 Chr.10: 50802424-50802424 on Build GRCh38
rs994263533 Chr.10: 50802166-50802166 on Build GRCh38
rs187640338 Chr.10: 50802045-50802045 on Build GRCh38
rs942452793 Chr.10: 50802514-50802514 on Build GRCh38
rs1038010437 Chr.10: 50802477-50802477 on Build GRCh38
rs74352101 Chr.10: 50802043-50802043 on Build GRCh38
rs1025762033 Chr.10: 50802172-50802172 on Build GRCh38
rs532026231 Chr.10: 50802384-50802384 on Build GRCh38
rs972262123 Chr.10: 50802042-50802042 on Build GRCh38
rs772148853 Chr.10: 50802413-50802413 on Build GRCh38
rs889722532 Chr.10: 50802394-50802394 on Build GRCh38
rs751812050 Chr.10: 50802128-50802128 on Build GRCh38
rs371949547 Chr.10: 50802319-50802319 on Build GRCh38
rs955165042 Chr.10: 50802404-50802404 on Build GRCh38
rs12571156 Chr.10: 50802159-50802159 on Build GRCh38
rs889438699 Chr.10: 50802082-50802082 on Build GRCh38
rs376351175 Chr.10: 50802332-50802332 on Build GRCh38
rs138030289 Chr.10: 50802340-50802340 on Build GRCh38
rs7072584 Chr.10: 50802069-50802069 on Build GRCh38
rs979860010 Chr.10: 50802313-50802313 on Build GRCh38
rs1006520441 Chr.10: 50802463-50802463 on Build GRCh38
rs747216594 Chr.10: 50802478-50802478 on Build GRCh38
rs545755151 Chr.10: 50802415-50802415 on Build GRCh38
rs755552918 Chr.10: 50802289-50802289 on Build GRCh38
rs371032681 Chr.10: 50802277-50802277 on Build GRCh38
rs892379711 Chr.10: 50802202-50802202 on Build GRCh38
rs375129305 Chr.10: 50802368-50802368 on Build GRCh38
rs570653825 Chr.10: 50802267-50802267 on Build GRCh38
rs183095006 Chr.10: 50802039-50802039 on Build GRCh38
rs769767018 Chr.10: 50802379-50802379 on Build GRCh38
rs988053238 Chr.10: 50802105-50802105 on Build GRCh38
rs550659290 Chr.10: 50802240-50802240 on Build GRCh38
rs781065153 Chr.10: 50802338-50802338 on Build GRCh38
rs150441974 Chr.10: 50802339-50802339 on Build GRCh38
rs192865589 Chr.10: 50802508-50802508 on Build GRCh38
rs549384923 Chr.10: 50802504-50802507 on Build GRCh38
rs992404765 Chr.10: 50802216-50802216 on Build GRCh38
rs916846076 Chr.10: 50802265-50802265 on Build GRCh38

View Full Product Details