The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199865.1 | NP_001186794.1 | 15 - 16 | 1771 | 113 |
NM_001199866.1 | NP_001186795.1 | 15 - 16 | 1771 | 113 | |
NM_024704.4 | NP_078980.3 | 15 - 16 | 1771 | 113 | |
XM_005260750.3 | XP_005260807.1 | 15 - 16 | 1750 | 113 | |
XM_005260751.4 | XP_005260808.1 | 15 - 16 | 1766 | 113 | |
XM_005260752.3 | XP_005260809.1 | 15 - 16 | 1750 | 113 | |
XM_005260753.3 | XP_005260810.1 | 15 - 16 | 1748 | 113 | |
XM_005260754.3 | XP_005260811.1 | 15 - 16 | 1748 | 113 | |
XM_005260755.3 | XP_005260812.1 | 15 - 16 | 1794 | 113 | |
XM_006723588.3 | XP_006723651.1 | 15 - 16 | 1748 | 113 | |
XM_017027926.1 | XP_016883415.1 | 15 - 16 | 1766 | 113 | |
GenBank mRNA | AB046810.2 | - | 15 - 16 | 1688 | 113 |
AK000142.1 | - | 9 - 10 | 1013 | 113 | |
AK095322.1 | - | 15 - 16 | 1708 | 113 | |
AK307399.1 | - | 15 - 16 | 1654 | 113 | |
AY166853.1 | - | 15 - 16 | 1770 | 113 | |
BC050577.1 | - | 15 - 16 | 1771 | 113 | |
BC150261.1 | - | 15 - 16 | 1688 | 113 | |
BX647572.1 | - | 15 - 16 | 1690 | 113 |