Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 60027679-60028191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANK3
Entrez Gene ID
288
Gene Name
ankyrin 3, node of Ranvier (ankyrin G)
Gene Aliases
ANKYRIN-G, MRT37
UniGene
Hs.499725
Gene Chromosome Location
Chr.10: 60026298-60733526 on Build GRCh38
Transcripts
NM_001149.3 NM_001204403.1 NM_001204404.1 NM_001320874.1 NM_020987.4

SNP Detail

SNP ID Chromosome Location
rs12265962 Chr.10: 60027793-60027793 on Build GRCh38
rs768981590 Chr.10: 60027743-60027743 on Build GRCh38
rs960108486 Chr.10: 60027910-60027910 on Build GRCh38
rs567125176 Chr.10: 60027731-60027731 on Build GRCh38
rs1057146185 Chr.10: 60027716-60027716 on Build GRCh38
rs996885221 Chr.10: 60027868-60027868 on Build GRCh38
rs901255300 Chr.10: 60027865-60027865 on Build GRCh38
rs545440357 Chr.10: 60027870-60027870 on Build GRCh38
rs866240026 Chr.10: 60028038-60028038 on Build GRCh38
rs369258431 Chr.10: 60028107-60028107 on Build GRCh38
rs564906805 Chr.10: 60028063-60028063 on Build GRCh38
rs147016684 Chr.10: 60028169-60028169 on Build GRCh38
rs368482147 Chr.10: 60027787-60027787 on Build GRCh38
rs942849332 Chr.10: 60028056-60028056 on Build GRCh38
rs1041554040 Chr.10: 60027862-60027862 on Build GRCh38
rs1040875993 Chr.10: 60028146-60028146 on Build GRCh38
rs115866436 Chr.10: 60027890-60027890 on Build GRCh38
rs980410444 Chr.10: 60028052-60028052 on Build GRCh38
rs950412269 Chr.10: 60027821-60027821 on Build GRCh38
rs773199328 Chr.10: 60027721-60027721 on Build GRCh38
rs7078873 Chr.10: 60027829-60027829 on Build GRCh38
rs913222478 Chr.10: 60027953-60027953 on Build GRCh38
rs1004790879 Chr.10: 60027823-60027823 on Build GRCh38
rs980347478 Chr.10: 60027864-60027864 on Build GRCh38
rs992990718 Chr.10: 60027956-60027956 on Build GRCh38
rs190133850 Chr.10: 60027752-60027752 on Build GRCh38
rs1028295197 Chr.10: 60027888-60027888 on Build GRCh38
rs762336223 Chr.10: 60027753-60027753 on Build GRCh38
rs542857204 Chr.10: 60027840-60027840 on Build GRCh38
rs145766077 Chr.10: 60027760-60027761 on Build GRCh38
rs904662887 Chr.10: 60027755-60027755 on Build GRCh38
rs749699833 Chr.10: 60028038-60028042 on Build GRCh38
rs762355181 Chr.10: 60028129-60028129 on Build GRCh38
rs979014686 Chr.10: 60028012-60028012 on Build GRCh38
rs997256749 Chr.10: 60027800-60027800 on Build GRCh38
rs1029953900 Chr.10: 60027818-60027818 on Build GRCh38
rs1015778641 Chr.10: 60027918-60027918 on Build GRCh38
rs946050294 Chr.10: 60028003-60028003 on Build GRCh38
rs992935518 Chr.10: 60027940-60027940 on Build GRCh38
rs961875833 Chr.10: 60027945-60027945 on Build GRCh38
rs1054082690 Chr.10: 60028122-60028122 on Build GRCh38
rs926200751 Chr.10: 60028037-60028037 on Build GRCh38
rs79598579 Chr.10: 60027724-60027724 on Build GRCh38
rs963201906 Chr.10: 60027834-60027834 on Build GRCh38
rs372524922 Chr.10: 60028039-60028039 on Build GRCh38
rs531929927 Chr.10: 60028100-60028100 on Build GRCh38
rs900268536 Chr.10: 60027798-60027798 on Build GRCh38
rs4633409 Chr.10: 60027930-60027930 on Build GRCh38
rs945243149 Chr.10: 60028132-60028133 on Build GRCh38
rs1038742029 Chr.10: 60027797-60027797 on Build GRCh38
rs4567399 Chr.10: 60027912-60027912 on Build GRCh38
rs549353002 Chr.10: 60028156-60028156 on Build GRCh38
rs372014030 Chr.10: 60028110-60028110 on Build GRCh38
rs926251736 Chr.10: 60028082-60028082 on Build GRCh38
rs901391842 Chr.10: 60028138-60028138 on Build GRCh38
rs578127688 Chr.10: 60027744-60027744 on Build GRCh38

View Full Product Details