Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 60028488-60028993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANK3
Entrez Gene ID
288
Gene Name
ankyrin 3, node of Ranvier (ankyrin G)
Gene Aliases
ANKYRIN-G, MRT37
UniGene
Hs.499725
Gene Chromosome Location
Chr.10: 60026298-60733526 on Build GRCh38
Transcripts
NM_001149.3 NM_001204403.1 NM_001204404.1 NM_001320874.1 NM_020987.4 XM_005269715.3 XM_006717796.3 XM_006717802.3 XM_011539708.2 XM_011539709.2 XM_017016110.1 XM_017016111.1 XM_017016112.1 XM_017016113.1 XM_017016114.1 XM_017016115.1 XM_017016116.1 XM_017016117.1 XM_017016118.1 XM_017016119.1 XM_017016120.1 XM_017016121.1 XM_017016122.1 XM_017016123.1 XM_017016124.1 XM_017016125.1 XM_017016126.1 XM_017016127.1 XM_017016128.1 XM_017016129.1 XM_017016130.1 XM_017016131.1 XM_017016132.1 XM_017016133.1 XM_017016134.1 XM_017016135.1 XM_017016136.1 XM_017016137.1 XM_017016138.1 XM_017016139.1 XM_017016140.1 XM_017016141.1 XM_017016142.1 XM_017016143.1 XM_017016144.1 XM_017016145.1 XM_017016146.1 XM_017016147.1 XM_017016148.1 XM_017016149.1 XM_017016150.1

SNP Detail

SNP ID Chromosome Location
rs1020335211 Chr.10: 60028665-60028665 on Build GRCh38
rs529783480 Chr.10: 60028895-60028895 on Build GRCh38
rs78055984 Chr.10: 60028642-60028642 on Build GRCh38
rs898855157 Chr.10: 60028536-60028536 on Build GRCh38
rs1020580660 Chr.10: 60028799-60028799 on Build GRCh38
rs115065245 Chr.10: 60028611-60028611 on Build GRCh38
rs1802665 Chr.10: 60028865-60028865 on Build GRCh38
rs548343084 Chr.10: 60028909-60028909 on Build GRCh38
rs897406896 Chr.10: 60028729-60028729 on Build GRCh38
rs908746297 Chr.10: 60028848-60028848 on Build GRCh38
rs572464747 Chr.10: 60028837-60028837 on Build GRCh38
rs932499475 Chr.10: 60028846-60028846 on Build GRCh38
rs187326819 Chr.10: 60028858-60028859 on Build GRCh38
rs897129253 Chr.10: 60028951-60028951 on Build GRCh38
rs976669317 Chr.10: 60028819-60028819 on Build GRCh38
rs150146053 Chr.10: 60028630-60028630 on Build GRCh38
rs906010749 Chr.10: 60028745-60028745 on Build GRCh38
rs199763470 Chr.10: 60028867-60028867 on Build GRCh38
rs895744371 Chr.10: 60028943-60028943 on Build GRCh38
rs955503185 Chr.10: 60028824-60028824 on Build GRCh38
rs1037025101 Chr.10: 60028722-60028722 on Build GRCh38
rs967859325 Chr.10: 60028743-60028743 on Build GRCh38
rs1014467908 Chr.10: 60028739-60028739 on Build GRCh38
rs778243921 Chr.10: 60028834-60028834 on Build GRCh38
rs370916512 Chr.10: 60028868-60028868 on Build GRCh38
rs72816487 Chr.10: 60028754-60028754 on Build GRCh38
rs11553751 Chr.10: 60028652-60028652 on Build GRCh38
rs964107635 Chr.10: 60028793-60028793 on Build GRCh38
rs1055682148 Chr.10: 60028936-60028936 on Build GRCh38
rs558928110 Chr.10: 60028725-60028725 on Build GRCh38
rs1049870992 Chr.10: 60028632-60028632 on Build GRCh38
rs941378697 Chr.10: 60028662-60028662 on Build GRCh38
rs989559533 Chr.10: 60028798-60028798 on Build GRCh38
rs369152683 Chr.10: 60028537-60028537 on Build GRCh38
rs1001439456 Chr.10: 60028544-60028544 on Build GRCh38
rs756668686 Chr.10: 60028815-60028815 on Build GRCh38
rs564532809 Chr.10: 60028859-60028859 on Build GRCh38
rs922876368 Chr.10: 60028817-60028817 on Build GRCh38
rs957776508 Chr.10: 60028788-60028788 on Build GRCh38
rs1037483537 Chr.10: 60028852-60028852 on Build GRCh38
rs1034280732 Chr.10: 60028569-60028569 on Build GRCh38
rs888516176 Chr.10: 60028633-60028633 on Build GRCh38
rs770341345 Chr.10: 60028913-60028913 on Build GRCh38
rs539043844 Chr.10: 60028602-60028602 on Build GRCh38
rs543755040 Chr.10: 60028849-60028849 on Build GRCh38
rs386744414 Chr.10: 60028867-60028868 on Build GRCh38
rs185539081 Chr.10: 60028784-60028784 on Build GRCh38
rs750576855 Chr.10: 60028516-60028516 on Build GRCh38

View Full Product Details