Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 61690275-61690806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf107
Entrez Gene ID
219621
Gene Name
chromosome 10 open reading frame 107
Gene Aliases
bA63A2.1
UniGene
Hs.673160
Gene Chromosome Location
Chr.10: 61662927-61766766 on Build GRCh38
Transcripts
NM_173554.2 XM_005269598.3 XM_005269599.4 XM_005269600.4 XM_006717690.3 XM_006717692.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4899925 c.285T>G p.T95T Chr.10: 61690618-61690618 on Build GRCh38
4482167 c.257C>T p.S86F Chr.10: 61690590-61690590 on Build GRCh38
6066706 c.223C>G p.Q75E Chr.10: 61690556-61690556 on Build GRCh38
7221365 c.288G>A p.E96E Chr.10: 61690621-61690621 on Build GRCh38
231211 c.223C>T p.Q75* Chr.10: 61690556-61690556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754781919 Chr.10: 61690527-61690527 on Build GRCh38
rs969597978 Chr.10: 61690748-61690748 on Build GRCh38
rs754239887 Chr.10: 61690567-61690567 on Build GRCh38
rs770873852 Chr.10: 61690619-61690619 on Build GRCh38
rs867463547 Chr.10: 61690437-61690437 on Build GRCh38
rs946340587 Chr.10: 61690533-61690533 on Build GRCh38
rs549481734 Chr.10: 61690633-61690633 on Build GRCh38
rs140653838 Chr.10: 61690573-61690573 on Build GRCh38
rs763663927 Chr.10: 61690565-61690565 on Build GRCh38
rs567643145 Chr.10: 61690402-61690402 on Build GRCh38
rs377640033 Chr.10: 61690530-61690530 on Build GRCh38
rs980875955 Chr.10: 61690497-61690497 on Build GRCh38
rs765889040 Chr.10: 61690390-61690390 on Build GRCh38
rs763344338 Chr.10: 61690672-61690672 on Build GRCh38
rs1016950697 Chr.10: 61690742-61690742 on Build GRCh38
rs748248447 Chr.10: 61690613-61690613 on Build GRCh38
rs572107180 Chr.10: 61690583-61690583 on Build GRCh38
rs761269755 Chr.10: 61690669-61690669 on Build GRCh38
rs1056745897 Chr.10: 61690388-61690388 on Build GRCh38
rs908305769 Chr.10: 61690373-61690373 on Build GRCh38
rs889046482 Chr.10: 61690370-61690370 on Build GRCh38
rs774195365 Chr.10: 61690531-61690531 on Build GRCh38
rs868541157 Chr.10: 61690556-61690556 on Build GRCh38
rs759370089 Chr.10: 61690532-61690532 on Build GRCh38
rs762368520 Chr.10: 61690570-61690570 on Build GRCh38
rs559532216 Chr.10: 61690326-61690326 on Build GRCh38
rs76616530 Chr.10: 61690517-61690517 on Build GRCh38
rs114060974 Chr.10: 61690563-61690563 on Build GRCh38
rs878992322 Chr.10: 61690754-61690754 on Build GRCh38
rs372325682 Chr.10: 61690321-61690321 on Build GRCh38
rs775144043 Chr.10: 61690554-61690554 on Build GRCh38
rs181105668 Chr.10: 61690376-61690376 on Build GRCh38
rs771882474 Chr.10: 61690622-61690622 on Build GRCh38
rs965399120 Chr.10: 61690504-61690504 on Build GRCh38
rs751837382 Chr.10: 61690599-61690599 on Build GRCh38
rs576439083 Chr.10: 61690749-61690749 on Build GRCh38
rs776200051 Chr.10: 61690644-61690644 on Build GRCh38
rs781389448 Chr.10: 61690761-61690761 on Build GRCh38
rs1018807805 Chr.10: 61690389-61690389 on Build GRCh38
rs935875224 Chr.10: 61690727-61690727 on Build GRCh38
rs866565042 Chr.10: 61690628-61690628 on Build GRCh38
rs939083361 Chr.10: 61690521-61690521 on Build GRCh38
rs916938513 Chr.10: 61690640-61690640 on Build GRCh38
rs768248871 Chr.10: 61690634-61690634 on Build GRCh38
rs1026118732 Chr.10: 61690661-61690661 on Build GRCh38
rs533058660 Chr.10: 61690482-61690482 on Build GRCh38
rs984148890 Chr.10: 61690452-61690452 on Build GRCh38
rs925900485 Chr.10: 61690700-61690700 on Build GRCh38
rs111272461 Chr.10: 61690671-61690671 on Build GRCh38
rs1044068390 Chr.10: 61690333-61690333 on Build GRCh38
rs1019654545 Chr.10: 61690501-61690501 on Build GRCh38
rs775054047 Chr.10: 61690625-61690625 on Build GRCh38
rs771686408 Chr.10: 61690540-61690540 on Build GRCh38
rs913602486 Chr.10: 61690778-61690778 on Build GRCh38
rs186135248 Chr.10: 61690768-61690768 on Build GRCh38
rs778867368 Chr.10: 61690620-61690620 on Build GRCh38
rs111291226 Chr.10: 61690558-61690558 on Build GRCh38
rs951497976 Chr.10: 61690443-61690443 on Build GRCh38
rs892568421 Chr.10: 61690497-61690497 on Build GRCh38
rs35796382 Chr.10: 61690621-61690621 on Build GRCh38
rs554880730 Chr.10: 61690627-61690627 on Build GRCh38
rs1005943820 Chr.10: 61690724-61690724 on Build GRCh38
rs374393780 Chr.10: 61690587-61690587 on Build GRCh38
rs891620163 Chr.10: 61690756-61690756 on Build GRCh38
rs910087595 Chr.10: 61690471-61690471 on Build GRCh38
rs116054413 Chr.10: 61690315-61690315 on Build GRCh38
rs560595319 Chr.10: 61690415-61690415 on Build GRCh38
rs781321991 Chr.10: 61690610-61690610 on Build GRCh38
rs768889226 Chr.10: 61690500-61690500 on Build GRCh38
rs754958507 Chr.10: 61690605-61690605 on Build GRCh38
rs1025641955 Chr.10: 61690417-61690417 on Build GRCh38
rs1041131751 Chr.10: 61690773-61690773 on Build GRCh38
rs1044414451 Chr.10: 61690692-61690692 on Build GRCh38

View Full Product Details