Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 61766215-61766764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf107
Entrez Gene ID
219621
Gene Name
chromosome 10 open reading frame 107
Gene Aliases
bA63A2.1
UniGene
Hs.673160
Gene Chromosome Location
Chr.10: 61662927-61766766 on Build GRCh38
Transcripts
NM_173554.2 XM_005269598.3 XM_005269599.4 XM_005269600.4 XM_006717690.3

SNP Detail

SNP ID Chromosome Location
rs1022881983 Chr.10: 61766302-61766302 on Build GRCh38
rs150963174 Chr.10: 61766507-61766507 on Build GRCh38
rs185895073 Chr.10: 61766613-61766613 on Build GRCh38
rs898340497 Chr.10: 61766276-61766276 on Build GRCh38
rs190183664 Chr.10: 61766359-61766359 on Build GRCh38
rs1048910113 Chr.10: 61766363-61766363 on Build GRCh38
rs942748822 Chr.10: 61766477-61766477 on Build GRCh38
rs985085460 Chr.10: 61766685-61766685 on Build GRCh38
rs985862986 Chr.10: 61766308-61766308 on Build GRCh38
rs906544927 Chr.10: 61766419-61766419 on Build GRCh38
rs974172882 Chr.10: 61766502-61766502 on Build GRCh38
rs1021824746 Chr.10: 61766363-61766369 on Build GRCh38
rs1022487880 Chr.10: 61766549-61766549 on Build GRCh38
rs1021369320 Chr.10: 61766619-61766619 on Build GRCh38
rs1011555074 Chr.10: 61766482-61766482 on Build GRCh38
rs61850530 Chr.10: 61766550-61766550 on Build GRCh38
rs965780240 Chr.10: 61766590-61766590 on Build GRCh38
rs528149470 Chr.10: 61766332-61766332 on Build GRCh38
rs948151816 Chr.10: 61766381-61766381 on Build GRCh38
rs1018673396 Chr.10: 61766566-61766566 on Build GRCh38
rs907091593 Chr.10: 61766556-61766557 on Build GRCh38
rs971983816 Chr.10: 61766341-61766341 on Build GRCh38
rs1011684053 Chr.10: 61766617-61766617 on Build GRCh38
rs182620997 Chr.10: 61766506-61766506 on Build GRCh38
rs756856680 Chr.10: 61766551-61766551 on Build GRCh38
rs971764253 Chr.10: 61766622-61766622 on Build GRCh38
rs957100373 Chr.10: 61766288-61766288 on Build GRCh38
rs537081315 Chr.10: 61766615-61766615 on Build GRCh38
rs754736611 Chr.10: 61766290-61766290 on Build GRCh38
rs930553739 Chr.10: 61766351-61766351 on Build GRCh38
rs397814668 Chr.10: 61766520-61766521 on Build GRCh38
rs1002460762 Chr.10: 61766310-61766310 on Build GRCh38
rs570579555 Chr.10: 61766242-61766242 on Build GRCh38
rs577288826 Chr.10: 61766641-61766641 on Build GRCh38
rs1038586703 Chr.10: 61766251-61766251 on Build GRCh38
rs929827203 Chr.10: 61766280-61766280 on Build GRCh38
rs752564951 Chr.10: 61766446-61766446 on Build GRCh38
rs558487401 Chr.10: 61766638-61766638 on Build GRCh38
rs911580649 Chr.10: 61766311-61766311 on Build GRCh38
rs187747456 Chr.10: 61766524-61766524 on Build GRCh38
rs978293523 Chr.10: 61766373-61766373 on Build GRCh38
rs11401832 Chr.10: 61766506-61766507 on Build GRCh38
rs181409382 Chr.10: 61766610-61766610 on Build GRCh38
rs955583738 Chr.10: 61766652-61766654 on Build GRCh38
rs964059238 Chr.10: 61766732-61766732 on Build GRCh38
rs1045104521 Chr.10: 61766387-61766387 on Build GRCh38
rs767413572 Chr.10: 61766303-61766303 on Build GRCh38
rs778538226 Chr.10: 61766645-61766645 on Build GRCh38
rs1018970930 Chr.10: 61766723-61766723 on Build GRCh38
rs918900712 Chr.10: 61766636-61766636 on Build GRCh38
rs1031172243 Chr.10: 61766265-61766265 on Build GRCh38
rs915764107 Chr.10: 61766718-61766718 on Build GRCh38
rs777119862 Chr.10: 61766534-61766534 on Build GRCh38
rs919216985 Chr.10: 61766346-61766346 on Build GRCh38
rs1031248798 Chr.10: 61766384-61766384 on Build GRCh38
rs758967989 Chr.10: 61766724-61766724 on Build GRCh38
rs79019245 Chr.10: 61766458-61766458 on Build GRCh38
rs1003768760 Chr.10: 61766240-61766240 on Build GRCh38
rs569992384 Chr.10: 61766556-61766556 on Build GRCh38
rs938620737 Chr.10: 61766565-61766565 on Build GRCh38
rs968227965 Chr.10: 61766372-61766372 on Build GRCh38
rs1033980544 Chr.10: 61766335-61766335 on Build GRCh38
rs1008496653 Chr.10: 61766681-61766681 on Build GRCh38
rs547974331 Chr.10: 61766338-61766338 on Build GRCh38
rs566102007 Chr.10: 61766367-61766367 on Build GRCh38
rs955614432 Chr.10: 61766408-61766408 on Build GRCh38
rs1003463996 Chr.10: 61766442-61766442 on Build GRCh38

View Full Product Details