Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 62804676-62805171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADO
Entrez Gene ID
84890
Gene Name
2-aminoethanethiol dioxygenase
Gene Aliases
C10orf22
UniGene
Hs.99821
Gene Chromosome Location
Chr.10: 62804756-62808479 on Build GRCh38
Transcripts
NM_032804.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416174 c.53G>A p.C18Y Chr.10: 62805112-62805112 on Build GRCh38
6420738 c.94G>T p.D32Y Chr.10: 62805153-62805153 on Build GRCh38
3397203 c.93C>T p.S31S Chr.10: 62805152-62805152 on Build GRCh38
6196973 c.93C>G p.S31S Chr.10: 62805152-62805152 on Build GRCh38
6582375 c.83G>T p.R28L Chr.10: 62805142-62805142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908773013 Chr.10: 62804803-62804803 on Build GRCh38
rs749792779 Chr.10: 62805112-62805112 on Build GRCh38
rs936477347 Chr.10: 62804956-62804956 on Build GRCh38
rs1026994997 Chr.10: 62804935-62804935 on Build GRCh38
rs763318320 Chr.10: 62805048-62805048 on Build GRCh38
rs906072898 Chr.10: 62804841-62804841 on Build GRCh38
rs765076270 Chr.10: 62804792-62804792 on Build GRCh38
rs987340228 Chr.10: 62804777-62804777 on Build GRCh38
rs1018735558 Chr.10: 62805109-62805109 on Build GRCh38
rs1000666380 Chr.10: 62804875-62804875 on Build GRCh38
rs1017162539 Chr.10: 62804861-62804861 on Build GRCh38
rs935965840 Chr.10: 62804926-62804926 on Build GRCh38
rs767725515 Chr.10: 62805127-62805127 on Build GRCh38
rs1056157857 Chr.10: 62804828-62804828 on Build GRCh38
rs891947651 Chr.10: 62804735-62804735 on Build GRCh38
rs553579183 Chr.10: 62804755-62804755 on Build GRCh38
rs377430884 Chr.10: 62805102-62805102 on Build GRCh38
rs775749855 Chr.10: 62805131-62805131 on Build GRCh38
rs1026458864 Chr.10: 62804881-62804881 on Build GRCh38
rs1052289471 Chr.10: 62805074-62805074 on Build GRCh38
rs770420193 Chr.10: 62805012-62805012 on Build GRCh38
rs758170027 Chr.10: 62805086-62805086 on Build GRCh38
rs964513504 Chr.10: 62805119-62805119 on Build GRCh38
rs373004863 Chr.10: 62804959-62804959 on Build GRCh38
rs1000823475 Chr.10: 62804739-62804739 on Build GRCh38
rs752746213 Chr.10: 62805064-62805064 on Build GRCh38
rs759989872 Chr.10: 62805126-62805126 on Build GRCh38
rs950499535 Chr.10: 62804884-62804884 on Build GRCh38
rs1000279857 Chr.10: 62804726-62804726 on Build GRCh38
rs950153528 Chr.10: 62804972-62804972 on Build GRCh38
rs747647989 Chr.10: 62805099-62805099 on Build GRCh38
rs1024956258 Chr.10: 62804746-62804746 on Build GRCh38
rs554379730 Chr.10: 62804860-62804860 on Build GRCh38
rs1020133192 Chr.10: 62804763-62804763 on Build GRCh38
rs1046271546 Chr.10: 62804996-62804996 on Build GRCh38
rs751307194 Chr.10: 62805144-62805144 on Build GRCh38
rs1053130117 Chr.10: 62804929-62804929 on Build GRCh38
rs1008178259 Chr.10: 62805090-62805090 on Build GRCh38
rs1008541918 Chr.10: 62804893-62804893 on Build GRCh38
rs986787762 Chr.10: 62804759-62804759 on Build GRCh38
rs773912961 Chr.10: 62805042-62805042 on Build GRCh38
rs1039061716 Chr.10: 62804711-62804711 on Build GRCh38
rs369703441 Chr.10: 62804788-62804788 on Build GRCh38
rs369730263 Chr.10: 62804892-62804892 on Build GRCh38
rs781057944 Chr.10: 62805098-62805098 on Build GRCh38
rs896128050 Chr.10: 62804853-62804853 on Build GRCh38
rs767776430 Chr.10: 62805062-62805062 on Build GRCh38
rs543492782 Chr.10: 62804889-62804889 on Build GRCh38
rs753893867 Chr.10: 62804865-62804865 on Build GRCh38
rs373580230 Chr.10: 62805097-62805097 on Build GRCh38
rs539288669 Chr.10: 62804748-62804748 on Build GRCh38
rs576043526 Chr.10: 62804863-62804863 on Build GRCh38
rs771278566 Chr.10: 62805124-62805124 on Build GRCh38
rs1033297217 Chr.10: 62804754-62804754 on Build GRCh38
rs913275460 Chr.10: 62804949-62804949 on Build GRCh38
rs906020578 Chr.10: 62805016-62805016 on Build GRCh38
rs557426887 Chr.10: 62804896-62804896 on Build GRCh38
rs929101698 Chr.10: 62804771-62804771 on Build GRCh38
rs754878156 Chr.10: 62805147-62805147 on Build GRCh38
rs199553864 Chr.10: 62805055-62805055 on Build GRCh38
rs764612485 Chr.10: 62804766-62804766 on Build GRCh38
rs764409805 Chr.10: 62804728-62804728 on Build GRCh38
rs531829189 Chr.10: 62805079-62805079 on Build GRCh38
rs545691421 Chr.10: 62804973-62804973 on Build GRCh38
rs146318332 Chr.10: 62804920-62804920 on Build GRCh38
rs936016668 Chr.10: 62804800-62804800 on Build GRCh38
rs549513740 Chr.10: 62805056-62805056 on Build GRCh38
rs778812081 Chr.10: 62805071-62805071 on Build GRCh38
rs1024487699 Chr.10: 62804831-62804831 on Build GRCh38
rs943259243 Chr.10: 62804790-62804790 on Build GRCh38
rs952897960 Chr.10: 62804938-62804938 on Build GRCh38
rs538530679 Chr.10: 62804753-62804753 on Build GRCh38
rs982889750 Chr.10: 62804940-62804940 on Build GRCh38
rs904506813 Chr.10: 62804799-62804799 on Build GRCh38
rs746866115 Chr.10: 62805093-62805093 on Build GRCh38
rs947864582 Chr.10: 62804862-62804862 on Build GRCh38
rs571249056 Chr.10: 62804747-62804747 on Build GRCh38
rs970288869 Chr.10: 62804749-62804749 on Build GRCh38
rs759700601 Chr.10: 62805059-62805059 on Build GRCh38
rs943208341 Chr.10: 62804904-62804904 on Build GRCh38
rs779732034 Chr.10: 62805089-62805089 on Build GRCh38
rs556090915 Chr.10: 62804717-62804717 on Build GRCh38
rs572168906 Chr.10: 62804773-62804773 on Build GRCh38
rs1001737413 Chr.10: 62804857-62804857 on Build GRCh38
rs564862856 Chr.10: 62804897-62804897 on Build GRCh38
rs975888145 Chr.10: 62804772-62804772 on Build GRCh38
rs761448815 Chr.10: 62805010-62805010 on Build GRCh38
rs1001685158 Chr.10: 62805035-62805035 on Build GRCh38
rs750322545 Chr.10: 62805137-62805137 on Build GRCh38
rs370370572 Chr.10: 62805067-62805067 on Build GRCh38
rs974645536 Chr.10: 62804906-62804906 on Build GRCh38
rs764188100 Chr.10: 62805134-62805134 on Build GRCh38
rs560737425 Chr.10: 62804977-62804977 on Build GRCh38
rs960349126 Chr.10: 62804941-62804941 on Build GRCh38
rs973107796 Chr.10: 62804758-62804758 on Build GRCh38
rs983005706 Chr.10: 62804802-62804802 on Build GRCh38
rs561226862 Chr.10: 62805070-62805070 on Build GRCh38
rs542775198 Chr.10: 62804822-62804822 on Build GRCh38
rs766375529 Chr.10: 62805143-62805143 on Build GRCh38
rs2236295 Chr.10: 62805132-62805132 on Build GRCh38
rs891899380 Chr.10: 62804810-62804810 on Build GRCh38
rs769440846 Chr.10: 62805101-62805101 on Build GRCh38
rs1014440114 Chr.10: 62804736-62804736 on Build GRCh38
rs1053586075 Chr.10: 62804808-62804808 on Build GRCh38
rs758243230 Chr.10: 62805141-62805141 on Build GRCh38

View Full Product Details