Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 62805075-62805589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADO
Entrez Gene ID
84890
Gene Name
2-aminoethanethiol dioxygenase
Gene Aliases
C10orf22
UniGene
Hs.99821
Gene Chromosome Location
Chr.10: 62804756-62808479 on Build GRCh38
Transcripts
NM_032804.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3686812 c.114C>T p.G38G Chr.10: 62805173-62805173 on Build GRCh38
6416174 c.53G>A p.C18Y Chr.10: 62805112-62805112 on Build GRCh38
3439552 c.280G>A p.D94N Chr.10: 62805339-62805339 on Build GRCh38
6420738 c.94G>T p.D32Y Chr.10: 62805153-62805153 on Build GRCh38
3397203 c.93C>T p.S31S Chr.10: 62805152-62805152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769921006 Chr.10: 62805292-62805292 on Build GRCh38
rs749792779 Chr.10: 62805112-62805112 on Build GRCh38
rs762381546 Chr.10: 62805562-62805562 on Build GRCh38
rs774993525 Chr.10: 62805168-62805168 on Build GRCh38
rs1018735558 Chr.10: 62805109-62805109 on Build GRCh38
rs771152092 Chr.10: 62805482-62805482 on Build GRCh38
rs780372448 Chr.10: 62805256-62805256 on Build GRCh38
rs748879301 Chr.10: 62805164-62805164 on Build GRCh38
rs767725515 Chr.10: 62805127-62805127 on Build GRCh38
rs377430884 Chr.10: 62805102-62805102 on Build GRCh38
rs775749855 Chr.10: 62805131-62805131 on Build GRCh38
rs201201168 Chr.10: 62805226-62805226 on Build GRCh38
rs760613955 Chr.10: 62805309-62805309 on Build GRCh38
rs374935591 Chr.10: 62805341-62805341 on Build GRCh38
rs772442170 Chr.10: 62805173-62805173 on Build GRCh38
rs571286399 Chr.10: 62805156-62805156 on Build GRCh38
rs752065369 Chr.10: 62805422-62805422 on Build GRCh38
rs964513504 Chr.10: 62805119-62805119 on Build GRCh38
rs781636658 Chr.10: 62805447-62805447 on Build GRCh38
rs769712637 Chr.10: 62805361-62805361 on Build GRCh38
rs776969451 Chr.10: 62805281-62805281 on Build GRCh38
rs759989872 Chr.10: 62805126-62805126 on Build GRCh38
rs775630587 Chr.10: 62805377-62805377 on Build GRCh38
rs747647989 Chr.10: 62805099-62805099 on Build GRCh38
rs748431849 Chr.10: 62805353-62805353 on Build GRCh38
rs746208878 Chr.10: 62805171-62805171 on Build GRCh38
rs757958121 Chr.10: 62805413-62805413 on Build GRCh38
rs371720949 Chr.10: 62805331-62805331 on Build GRCh38
rs778518893 Chr.10: 62805329-62805329 on Build GRCh38
rs780601560 Chr.10: 62805339-62805339 on Build GRCh38
rs145507280 Chr.10: 62805396-62805396 on Build GRCh38
rs754751741 Chr.10: 62805204-62805204 on Build GRCh38
rs751307194 Chr.10: 62805144-62805144 on Build GRCh38
rs772133073 Chr.10: 62805375-62805375 on Build GRCh38
rs763645049 Chr.10: 62805311-62805311 on Build GRCh38
rs937485716 Chr.10: 62805227-62805227 on Build GRCh38
rs764345807 Chr.10: 62805176-62805176 on Build GRCh38
rs758869157 Chr.10: 62805335-62805335 on Build GRCh38
rs560631386 Chr.10: 62805555-62805555 on Build GRCh38
rs766213047 Chr.10: 62805197-62805197 on Build GRCh38
rs1045800824 Chr.10: 62805209-62805209 on Build GRCh38
rs748512266 Chr.10: 62805452-62805452 on Build GRCh38
rs368304557 Chr.10: 62805249-62805249 on Build GRCh38
rs1040982637 Chr.10: 62805543-62805543 on Build GRCh38
rs781057944 Chr.10: 62805098-62805098 on Build GRCh38
rs755256834 Chr.10: 62805446-62805446 on Build GRCh38
rs771278566 Chr.10: 62805124-62805124 on Build GRCh38
rs373580230 Chr.10: 62805097-62805097 on Build GRCh38
rs753546467 Chr.10: 62805326-62805326 on Build GRCh38
rs777522209 Chr.10: 62805242-62805242 on Build GRCh38
rs781204073 Chr.10: 62805343-62805343 on Build GRCh38
rs762015561 Chr.10: 62805195-62805195 on Build GRCh38
rs760493176 Chr.10: 62805395-62805395 on Build GRCh38
rs540992216 Chr.10: 62805492-62805492 on Build GRCh38
rs754878156 Chr.10: 62805147-62805147 on Build GRCh38
rs10995311 Chr.10: 62805174-62805174 on Build GRCh38
rs768289642 Chr.10: 62805505-62805505 on Build GRCh38
rs1004241752 Chr.10: 62805248-62805248 on Build GRCh38
rs576932938 Chr.10: 62805489-62805489 on Build GRCh38
rs146642029 Chr.10: 62805243-62805243 on Build GRCh38
rs576095610 Chr.10: 62805390-62805390 on Build GRCh38
rs749366294 Chr.10: 62805472-62805472 on Build GRCh38
rs763957254 Chr.10: 62805401-62805401 on Build GRCh38
rs764835246 Chr.10: 62805406-62805406 on Build GRCh38
rs779233980 Chr.10: 62805246-62805246 on Build GRCh38
rs776538639 Chr.10: 62805181-62805181 on Build GRCh38
rs143503748 Chr.10: 62805416-62805416 on Build GRCh38
rs148838445 Chr.10: 62805407-62805407 on Build GRCh38
rs558611751 Chr.10: 62805485-62805485 on Build GRCh38
rs751527435 Chr.10: 62805203-62805203 on Build GRCh38
rs761740161 Chr.10: 62805283-62805283 on Build GRCh38
rs1008993745 Chr.10: 62805373-62805373 on Build GRCh38
rs756657888 Chr.10: 62805328-62805328 on Build GRCh38
rs538620100 Chr.10: 62805161-62805161 on Build GRCh38
rs374769704 Chr.10: 62805564-62805564 on Build GRCh38
rs371172315 Chr.10: 62805356-62805356 on Build GRCh38
rs763166960 Chr.10: 62805367-62805367 on Build GRCh38
rs61744064 Chr.10: 62805251-62805251 on Build GRCh38
rs971392296 Chr.10: 62805557-62805557 on Build GRCh38
rs113773165 Chr.10: 62805508-62805508 on Build GRCh38
rs752509629 Chr.10: 62805307-62805307 on Build GRCh38
rs773406137 Chr.10: 62805366-62805366 on Build GRCh38
rs943836969 Chr.10: 62805502-62805502 on Build GRCh38
rs759570832 Chr.10: 62805301-62805301 on Build GRCh38
rs756330689 Chr.10: 62805455-62805455 on Build GRCh38
rs767449964 Chr.10: 62805210-62805210 on Build GRCh38
rs369053424 Chr.10: 62805409-62805409 on Build GRCh38
rs768856931 Chr.10: 62805268-62805268 on Build GRCh38
rs750322545 Chr.10: 62805137-62805137 on Build GRCh38
rs372219241 Chr.10: 62805166-62805166 on Build GRCh38
rs867305564 Chr.10: 62805441-62805441 on Build GRCh38
rs753395175 Chr.10: 62805403-62805403 on Build GRCh38
rs574219082 Chr.10: 62805294-62805294 on Build GRCh38
rs778208521 Chr.10: 62805465-62805465 on Build GRCh38
rs764188100 Chr.10: 62805134-62805134 on Build GRCh38
rs755647923 Chr.10: 62805159-62805159 on Build GRCh38
rs747411415 Chr.10: 62805259-62805259 on Build GRCh38
rs761576780 Chr.10: 62805527-62805527 on Build GRCh38
rs374385777 Chr.10: 62805522-62805522 on Build GRCh38
rs541712569 Chr.10: 62805404-62805404 on Build GRCh38
rs867885321 Chr.10: 62805196-62805196 on Build GRCh38
rs202015367 Chr.10: 62805552-62805552 on Build GRCh38
rs755807283 Chr.10: 62805237-62805237 on Build GRCh38
rs766375529 Chr.10: 62805143-62805143 on Build GRCh38
rs2236295 Chr.10: 62805132-62805132 on Build GRCh38
rs145126961 Chr.10: 62805362-62805362 on Build GRCh38
rs769440846 Chr.10: 62805101-62805101 on Build GRCh38
rs767613495 Chr.10: 62805303-62805303 on Build GRCh38
rs760907121 Chr.10: 62805175-62805175 on Build GRCh38
rs565928292 Chr.10: 62805179-62805179 on Build GRCh38
rs776452324 Chr.10: 62805506-62805506 on Build GRCh38
rs758243230 Chr.10: 62805141-62805141 on Build GRCh38

View Full Product Details