Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 62806659-62807162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADO
Entrez Gene ID
84890
Gene Name
2-aminoethanethiol dioxygenase
Gene Aliases
C10orf22
UniGene
Hs.99821
Gene Chromosome Location
Chr.10: 62804756-62808479 on Build GRCh38
Transcripts
NM_032804.5

SNP Detail

SNP ID Chromosome Location
rs995824817 Chr.10: 62806826-62806826 on Build GRCh38
rs531044630 Chr.10: 62807097-62807097 on Build GRCh38
rs139346675 Chr.10: 62806708-62806708 on Build GRCh38
rs570643934 Chr.10: 62807028-62807028 on Build GRCh38
rs12779224 Chr.10: 62806791-62806791 on Build GRCh38
rs973062677 Chr.10: 62806928-62806928 on Build GRCh38
rs928405587 Chr.10: 62806762-62806762 on Build GRCh38
rs541490917 Chr.10: 62806873-62806873 on Build GRCh38
rs552440622 Chr.10: 62807024-62807024 on Build GRCh38
rs574697370 Chr.10: 62806731-62806731 on Build GRCh38
rs766166376 Chr.10: 62806917-62806917 on Build GRCh38
rs530119955 Chr.10: 62806908-62806908 on Build GRCh38
rs931188056 Chr.10: 62807076-62807076 on Build GRCh38
rs915724464 Chr.10: 62807069-62807069 on Build GRCh38
rs534696271 Chr.10: 62806703-62806703 on Build GRCh38
rs971823071 Chr.10: 62806943-62806946 on Build GRCh38
rs745560942 Chr.10: 62806807-62806807 on Build GRCh38
rs1028489653 Chr.10: 62806910-62806910 on Build GRCh38
rs1021236550 Chr.10: 62806926-62806926 on Build GRCh38
rs140167418 Chr.10: 62807079-62807079 on Build GRCh38
rs193201560 Chr.10: 62806951-62806951 on Build GRCh38
rs1019688957 Chr.10: 62806900-62806900 on Build GRCh38
rs149623368 Chr.10: 62806763-62806763 on Build GRCh38
rs185388793 Chr.10: 62806954-62806954 on Build GRCh38
rs1048329131 Chr.10: 62806872-62806872 on Build GRCh38
rs887093033 Chr.10: 62806896-62806896 on Build GRCh38
rs76511847 Chr.10: 62806770-62806770 on Build GRCh38
rs75351521 Chr.10: 62806902-62806902 on Build GRCh38
rs768667674 Chr.10: 62806697-62806697 on Build GRCh38
rs913407154 Chr.10: 62807065-62807065 on Build GRCh38
rs1048762681 Chr.10: 62807091-62807091 on Build GRCh38
rs189948695 Chr.10: 62806705-62806705 on Build GRCh38
rs950040111 Chr.10: 62806737-62806737 on Build GRCh38
rs775738319 Chr.10: 62807035-62807035 on Build GRCh38
rs765384863 Chr.10: 62806775-62806775 on Build GRCh38
rs1041830907 Chr.10: 62806752-62806752 on Build GRCh38
rs747109275 Chr.10: 62806749-62806749 on Build GRCh38
rs541815751 Chr.10: 62806933-62806933 on Build GRCh38
rs555254351 Chr.10: 62806740-62806740 on Build GRCh38
rs1005243835 Chr.10: 62806918-62806918 on Build GRCh38
rs144345885 Chr.10: 62806861-62806861 on Build GRCh38
rs10995312 Chr.10: 62806812-62806812 on Build GRCh38
rs1016757751 Chr.10: 62806919-62806919 on Build GRCh38
rs938425082 Chr.10: 62807000-62807000 on Build GRCh38
rs932205492 Chr.10: 62807129-62807129 on Build GRCh38
rs928352683 Chr.10: 62806973-62806973 on Build GRCh38
rs990121548 Chr.10: 62806924-62806924 on Build GRCh38
rs776681251 Chr.10: 62806736-62806736 on Build GRCh38
rs980123785 Chr.10: 62807003-62807003 on Build GRCh38
rs887050489 Chr.10: 62807112-62807112 on Build GRCh38
rs927266396 Chr.10: 62807008-62807008 on Build GRCh38
rs866423277 Chr.10: 62806878-62806878 on Build GRCh38
rs1004232850 Chr.10: 62806899-62806899 on Build GRCh38
rs1054544145 Chr.10: 62807047-62807047 on Build GRCh38
rs751497077 Chr.10: 62807013-62807013 on Build GRCh38
rs754926988 Chr.10: 62807058-62807058 on Build GRCh38
rs535072571 Chr.10: 62806710-62806710 on Build GRCh38
rs1041040818 Chr.10: 62807074-62807074 on Build GRCh38
rs989783228 Chr.10: 62806719-62806719 on Build GRCh38

View Full Product Details