Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEPACAM2
Entrez Gene ID
253012
Gene Name
HEPACAM family member 2
Gene Aliases
MIKI
Chromosome Location
Chr.7: 93188586 - 93232297 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039372.2 NP_001034461.1 8 - 9 1348 100
NM_001288804.1 NP_001275733.1 9 - 10 1417 100
NM_001288810.1 NP_001275739.1 6 - 7 1315 100
NM_198151.2 NP_937794.1 7 - 8 1340 100
XM_011516000.2 XP_011514302.1 8 - 9 1281 100
XM_011516001.1 XP_011514303.1 8 - 9 1281 100
GenBank mRNA AK096002.1 - 8 - 9 1301 100
AK298361.1 - 9 - 10 1367 100
BC136300.1 - 7 - 8 1270 100
BC136301.1 - 7 - 8 1270 100
BC139906.1 - 7 - 8 1327 100

View Full Product Details