The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172630.1 | NP_001166101.1 | 13 - 14 | 1356 | 70 |
NM_052948.3 | NP_443180.2 | 14 - 15 | 1454 | 70 | |
XM_005258488.1 | XP_005258545.1 | 14 - 15 | 1450 | 70 | |
XM_006722999.1 | XP_006723062.1 | 14 - 15 | 1453 | 70 | |
XM_011526416.1 | XP_011524718.1 | 13 - 14 | 1308 | 70 | |
XM_011526417.1 | XP_011524719.1 | 13 - 14 | 1311 | 70 | |
XM_011526418.1 | XP_011524720.1 | 13 - 14 | 1311 | 70 | |
XM_011526419.1 | XP_011524721.1 | 13 - 14 | 1308 | 70 | |
XM_011526420.1 | XP_011524722.1 | 13 - 14 | 1359 | 70 | |
XM_011526421.2 | XP_011524723.1 | 13 - 14 | 2853 | 70 | |
XM_017026238.1 | XP_016881727.1 | 13 - 14 | 2853 | 70 | |
GenBank mRNA | AK096338.1 | - | 14 - 15 | 1395 | 70 |
AK127255.1 | - | 13 - 14 | 1356 | 70 | |
BC128586.1 | - | 14 - 15 | 1367 | 70 |