The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172630.1 | NP_001166101.1 | 14 - 15 | 1462 | 71 |
NM_052948.3 | NP_443180.2 | 15 - 16 | 1560 | 71 | |
XM_005258488.1 | XP_005258545.1 | 15 - 16 | 1556 | 71 | |
XM_006722999.1 | XP_006723062.1 | 15 - 16 | 1559 | 71 | |
XM_011526416.1 | XP_011524718.1 | 14 - 15 | 1414 | 71 | |
XM_011526417.1 | XP_011524719.1 | 14 - 15 | 1417 | 71 | |
XM_011526418.1 | XP_011524720.1 | 14 - 15 | 1417 | 71 | |
XM_011526419.1 | XP_011524721.1 | 14 - 15 | 1414 | 71 | |
XM_011526420.1 | XP_011524722.1 | 14 - 15 | 1465 | 71 | |
XM_011526421.2 | XP_011524723.1 | 14 - 15 | 2959 | 71 | |
XM_017026238.1 | XP_016881727.1 | 14 - 15 | 2959 | 71 | |
GenBank mRNA | AK096338.1 | - | 15 - 16 | 1501 | 71 |
AK127255.1 | - | 14 - 15 | 1462 | 71 | |
AL137579.1 | - | 2 - 3 | 142 | 71 | |
BC128586.1 | - | 15 - 16 | 1473 | 71 |