Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 6873843-6874345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMELY
Entrez Gene ID
266
Gene Name
amelogenin, Y-linked
Gene Aliases
AMGL, AMGY
UniGene
Hs.1238
Gene Chromosome Location
Chr.Y: 6865918-6874056 on Build GRCh38
Transcripts
NM_001143.1 XM_017030042.1

SNP Detail

SNP ID Chromosome Location
rs866886131 Chr.Y: 6874238-6874238 on Build GRCh38
rs774722804 Chr.Y: 6874167-6874167 on Build GRCh38
rs1020823061 Chr.Y: 6874037-6874037 on Build GRCh38
rs764004843 Chr.Y: 6874088-6874088 on Build GRCh38
rs979325877 Chr.Y: 6874059-6874059 on Build GRCh38
rs374782962 Chr.Y: 6874156-6874156 on Build GRCh38
rs751503948 Chr.Y: 6873971-6873971 on Build GRCh38
rs966630006 Chr.Y: 6874039-6874039 on Build GRCh38

View Full Product Details