Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OIT3
Entrez Gene ID
170392
Gene Name
oncoprotein induced transcript 3
Gene Aliases
LZP
Chromosome Location
Chr.10: 72893556 - 72933036 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152635.2 NP_689848.1 7 - 8 1609 71
GenBank mRNA AK096435.1 - 7 - 8 1584 71
AY013707.2 - 7 - 8 1440 71
AY358339.1 - 7 - 8 1562 71

View Full Product Details