The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173483.3 | NP_775754.2 | 10 - 11 | 1298 | 81 |
XM_011527692.2 | XP_011525994.1 | 11 - 12 | 1540 | 81 | |
XM_011527693.2 | XP_011525995.1 | 10 - 11 | 1310 | 81 | |
GenBank mRNA | AK096820.1 | - | 10 - 11 | 1298 | 81 |
BC069351.1 | - | 9 - 10 | 1235 | 81 | |
BC093894.1 | - | 9 - 10 | 1172 | 81 | |
BC093896.1 | - | 9 - 10 | 1172 | 81 |