Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPM1N
Entrez Gene ID
147699
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1N (putative)
Gene Aliases
-
Chromosome Location
Chr.19: 45497964 - 45502694 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080401.1 NP_001073870.1 4 - 5 1226 114
GenBank mRNA AK097444.1 - 3 - 4 1629 114
BC062452.1 - 4 - 5 404 114

View Full Product Details