Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR93
Entrez Gene ID
56964
Gene Name
WD repeat domain 93
Gene Aliases
-
Chromosome Location
Chr.15: 89690074 - 89744390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284395.1 NP_001271324.1 14 - 15 1627 58
NM_020212.1 NP_064597.1 14 - 15 1711 58
GenBank mRNA AK097525.1 - 14 - 15 1595 58
AK302032.1 - 8 - 9 1116 58
AL390084.1 - 6 - 7 695 58
AL390085.1 - 6 - 7 695 58
BC064626.1 - 14 - 15 1711 58

View Full Product Details