Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1044543-1045087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA7
Entrez Gene ID
10347
Gene Name
ATP binding cassette subfamily A member 7
Gene Aliases
ABCA-SSN, ABCX, AD9
UniGene
Hs.134514
Gene Chromosome Location
Chr.19: 1040103-1065572 on Build GRCh38
Transcripts
NM_019112.3 XM_006722616.1 XM_006722617.2 XM_011527628.2 XM_011527629.1 XM_011527630.1 XM_011527631.1 XM_011527632.1 XM_011527633.2 XM_011527634.1 XM_011527635.1 XM_017026142.1 XM_017026143.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3970668 c.1164C>T p.D388D Chr.19: 1044693-1044693 on Build GRCh38
1480515 c.1250C>G p.S417* Chr.19: 1045036-1045036 on Build GRCh38
3388541 c.1088G>T p.R363I Chr.19: 1044617-1044617 on Build GRCh38
6589828 c.1298G>A p.R433Q Chr.19: 1045084-1045084 on Build GRCh38
474105 c.1171G>A p.A391T Chr.19: 1044700-1044700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771364442 Chr.19: 1044645-1044645 on Build GRCh38
rs953441463 Chr.19: 1044826-1044826 on Build GRCh38
rs376514872 Chr.19: 1044691-1044691 on Build GRCh38
rs763954501 Chr.19: 1044986-1044986 on Build GRCh38
rs768822779 Chr.19: 1044717-1044717 on Build GRCh38
rs373349507 Chr.19: 1044612-1044612 on Build GRCh38
rs142291535 Chr.19: 1045056-1045056 on Build GRCh38
rs770896698 Chr.19: 1044959-1044959 on Build GRCh38
rs780368480 Chr.19: 1044767-1044767 on Build GRCh38
rs35732553 Chr.19: 1044700-1044701 on Build GRCh38
rs777976642 Chr.19: 1045005-1045005 on Build GRCh38
rs779016991 Chr.19: 1045001-1045001 on Build GRCh38
rs1018582403 Chr.19: 1044996-1044996 on Build GRCh38
rs3764648 Chr.19: 1044754-1044754 on Build GRCh38
rs574102212 Chr.19: 1044782-1044782 on Build GRCh38
rs761274831 Chr.19: 1045044-1045044 on Build GRCh38
rs757137311 Chr.19: 1044615-1044615 on Build GRCh38
rs879085197 Chr.19: 1044999-1044999 on Build GRCh38
rs776835870 Chr.19: 1044646-1044646 on Build GRCh38
rs149365975 Chr.19: 1044705-1044705 on Build GRCh38
rs754600576 Chr.19: 1044693-1044693 on Build GRCh38
rs1024172178 Chr.19: 1044813-1044813 on Build GRCh38
rs765604779 Chr.19: 1045057-1045057 on Build GRCh38
rs762984064 Chr.19: 1044609-1044609 on Build GRCh38
rs749509977 Chr.19: 1044714-1044714 on Build GRCh38
rs747465627 Chr.19: 1044638-1044638 on Build GRCh38
rs778252749 Chr.19: 1044703-1044703 on Build GRCh38
rs749562991 Chr.19: 1044772-1044772 on Build GRCh38
rs750972264 Chr.19: 1044984-1044984 on Build GRCh38
rs766877133 Chr.19: 1045047-1045047 on Build GRCh38
rs372454069 Chr.19: 1044923-1044923 on Build GRCh38
rs752176682 Chr.19: 1044752-1044752 on Build GRCh38
rs767395449 Chr.19: 1044660-1044660 on Build GRCh38
rs878915585 Chr.19: 1044891-1044891 on Build GRCh38
rs765352552 Chr.19: 1044596-1044596 on Build GRCh38
rs752910380 Chr.19: 1045066-1045066 on Build GRCh38
rs1034222817 Chr.19: 1045034-1045034 on Build GRCh38
rs761627515 Chr.19: 1044659-1044659 on Build GRCh38
rs764892701 Chr.19: 1044751-1044751 on Build GRCh38
rs776588492 Chr.19: 1044964-1044964 on Build GRCh38
rs199847666 Chr.19: 1044982-1044982 on Build GRCh38
rs745659364 Chr.19: 1045026-1045026 on Build GRCh38
rs776344453 Chr.19: 1044568-1044569 on Build GRCh38
rs148078867 Chr.19: 1044727-1044727 on Build GRCh38
rs10405305 Chr.19: 1045027-1045027 on Build GRCh38
rs776981452 Chr.19: 1044587-1044587 on Build GRCh38
rs924752657 Chr.19: 1044844-1044844 on Build GRCh38
rs999911593 Chr.19: 1044745-1044745 on Build GRCh38
rs748181239 Chr.19: 1044787-1044787 on Build GRCh38
rs372269262 Chr.19: 1045038-1045038 on Build GRCh38
rs944900994 Chr.19: 1044942-1044942 on Build GRCh38
rs547842844 Chr.19: 1044840-1044840 on Build GRCh38
rs746230971 Chr.19: 1044710-1044710 on Build GRCh38
rs577493277 Chr.19: 1044637-1044637 on Build GRCh38
rs754313265 Chr.19: 1045049-1045049 on Build GRCh38
rs759332354 Chr.19: 1044972-1044972 on Build GRCh38
rs755304837 Chr.19: 1044998-1044998 on Build GRCh38
rs764070811 Chr.19: 1044653-1044653 on Build GRCh38
rs748713198 Chr.19: 1044569-1044569 on Build GRCh38
rs367921437 Chr.19: 1044888-1044888 on Build GRCh38
rs990754334 Chr.19: 1044912-1044912 on Build GRCh38
rs138055574 Chr.19: 1044673-1044673 on Build GRCh38
rs774415633 Chr.19: 1044655-1044655 on Build GRCh38
rs527445986 Chr.19: 1044843-1044843 on Build GRCh38
rs3764647 Chr.19: 1044713-1044713 on Build GRCh38
rs762638418 Chr.19: 1045028-1045028 on Build GRCh38
rs971693640 Chr.19: 1044823-1044823 on Build GRCh38
rs146070541 Chr.19: 1045019-1045019 on Build GRCh38
rs774275278 Chr.19: 1044718-1044718 on Build GRCh38
rs770030201 Chr.19: 1044649-1044649 on Build GRCh38
rs757724125 Chr.19: 1044708-1044708 on Build GRCh38
rs1027559028 Chr.19: 1044818-1044818 on Build GRCh38
rs983996048 Chr.19: 1044921-1044921 on Build GRCh38
rs754187080 Chr.19: 1044991-1044991 on Build GRCh38
rs760603867 Chr.19: 1044734-1044734 on Build GRCh38
rs767214073 Chr.19: 1044619-1044619 on Build GRCh38
rs980009332 Chr.19: 1044839-1044839 on Build GRCh38
rs1018657381 Chr.19: 1044833-1044833 on Build GRCh38
rs773348955 Chr.19: 1044796-1044796 on Build GRCh38
rs771187987 Chr.19: 1044577-1044577 on Build GRCh38
rs531555427 Chr.19: 1044943-1044943 on Build GRCh38
rs766272429 Chr.19: 1044738-1044738 on Build GRCh38
rs759086782 Chr.19: 1044750-1044750 on Build GRCh38
rs142777510 Chr.19: 1045009-1045009 on Build GRCh38
rs544191282 Chr.19: 1044651-1044651 on Build GRCh38
rs567222111 Chr.19: 1044709-1044719 on Build GRCh38
rs552120252 Chr.19: 1044863-1044863 on Build GRCh38
rs372641327 Chr.19: 1044955-1044955 on Build GRCh38
rs566988992 Chr.19: 1045007-1045007 on Build GRCh38
rs751607002 Chr.19: 1044650-1044650 on Build GRCh38
rs143296441 Chr.19: 1044590-1044590 on Build GRCh38
rs928640181 Chr.19: 1044924-1044924 on Build GRCh38
rs747035056 Chr.19: 1045010-1045010 on Build GRCh38
rs750266608 Chr.19: 1044661-1044661 on Build GRCh38
rs779804502 Chr.19: 1044628-1044628 on Build GRCh38
rs772728537 Chr.19: 1044573-1044573 on Build GRCh38
rs750920945 Chr.19: 1044757-1044757 on Build GRCh38
rs773875632 Chr.19: 1045035-1045035 on Build GRCh38
rs916161554 Chr.19: 1044920-1044920 on Build GRCh38
rs751398887 Chr.19: 1044614-1044614 on Build GRCh38
rs779677941 Chr.19: 1044568-1044568 on Build GRCh38
rs746179458 Chr.19: 1044647-1044647 on Build GRCh38
rs955853075 Chr.19: 1044755-1044755 on Build GRCh38
rs558930994 Chr.19: 1045053-1045053 on Build GRCh38
rs766142894 Chr.19: 1044684-1044684 on Build GRCh38
rs772070834 Chr.19: 1044947-1044947 on Build GRCh38
rs370899120 Chr.19: 1044989-1044989 on Build GRCh38
rs773111881 Chr.19: 1044771-1044774 on Build GRCh38
rs775573683 Chr.19: 1044600-1044600 on Build GRCh38
rs368842046 Chr.19: 1044986-1044986 on Build GRCh38
rs764798425 Chr.19: 1044973-1044973 on Build GRCh38
rs35815587 Chr.19: 1044742-1044743 on Build GRCh38
rs757372417 Chr.19: 1044854-1044854 on Build GRCh38
rs3764649 Chr.19: 1044976-1044976 on Build GRCh38
rs781692277 Chr.19: 1044709-1044709 on Build GRCh38
rs762768623 Chr.19: 1044968-1044985 on Build GRCh38
rs761115996 Chr.19: 1044575-1044575 on Build GRCh38
rs973410134 Chr.19: 1044850-1044850 on Build GRCh38
rs765063029 Chr.19: 1044629-1044630 on Build GRCh38
rs772177954 Chr.19: 1044793-1044793 on Build GRCh38
rs771935858 Chr.19: 1044733-1044733 on Build GRCh38
rs541477795 Chr.19: 1044828-1044828 on Build GRCh38
rs940085221 Chr.19: 1044934-1044934 on Build GRCh38
rs371536711 Chr.19: 1044979-1044979 on Build GRCh38
rs146982710 Chr.19: 1044620-1044620 on Build GRCh38
rs753583915 Chr.19: 1044741-1044741 on Build GRCh38
rs756540366 Chr.19: 1044764-1044764 on Build GRCh38
rs768784238 Chr.19: 1044775-1044775 on Build GRCh38
rs1004259814 Chr.19: 1044734-1044735 on Build GRCh38
rs771021175 Chr.19: 1045016-1045016 on Build GRCh38
rs374214649 Chr.19: 1045003-1045003 on Build GRCh38
rs971361106 Chr.19: 1044578-1044578 on Build GRCh38
rs748838063 Chr.19: 1044630-1044630 on Build GRCh38
rs759154525 Chr.19: 1044569-1044569 on Build GRCh38
rs754479722 Chr.19: 1044631-1044631 on Build GRCh38

View Full Product Details