The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032290.3 | NP_115666.2 | 3 - 4 | 266 | 109 |
XM_017009978.1 | XP_016865467.1 | 3 - 4 | 447 | 109 | |
XM_017009979.1 | XP_016865468.1 | 3 - 4 | 5028 | 109 | |
XM_017009980.1 | XP_016865469.1 | 3 - 4 | 448 | 109 | |
XM_017009981.1 | XP_016865470.1 | 3 - 4 | 298 | 109 | |
XM_017009982.1 | XP_016865471.1 | 3 - 4 | 447 | 109 | |
GenBank mRNA | AK297452.1 | - | 3 - 4 | 274 | 109 |
AK299080.1 | - | 3 - 4 | 292 | 109 | |
AL050298.1 | - | 3 - 4 | 271 | 109 | |
BC070332.1 | - | 3 - 4 | 304 | 109 | |
BC107571.1 | - | 3 - 4 | 266 | 109 | |
CR936768.1 | - | 3 - 4 | 272 | 109 | |
DR001089.1 | - | 3 - 4 | 266 | 109 |