Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KDM2B
Entrez Gene ID
84678
Gene Name
lysine demethylase 2B
Gene Aliases
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2
Chromosome Location
Chr.12: 121402289 - 121582268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005366.1 NP_001005366.1 15 - 16 2507 60
NM_032590.4 NP_115979.3 15 - 16 2297 60
XM_005253955.3 XP_005254012.1 15 - 16 2507 60
XM_005253956.3 XP_005254013.1 15 - 16 2104 60
XM_005253961.4 XP_005254018.1 4 - 5 592 60
GenBank mRNA AK027440.1 - 5 - 6 686 60
AK027692.1 - 2 - 3 202 60
AK127328.1 - 15 - 16 2507 60
AK316470.1 - 4 - 5 604 60
AL133572.1 - 10 - 11 2548 60
BC008735.2 - 1 - 2 67 60

View Full Product Details